missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ opus™Titrator

Product Code. 10657774
Change view
Click to view available options
Volume (Metric) Sample:
50 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sample Unit Size
10657774 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10657774 Supplier Hirschmann™ Supplier No. 9582050
  • ONLINE EXCLUSIVE PRICE
    2665.00 €
    1924.00 € / Pack of 1
    Save 741.00 €
    27% Off
View all promotions
Estimated Shipment: 19-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Titration unit with a reagent recirculation system for motor-powered titration directly from the reagent bottle.

  • Electronic control and dispensing of volumes
  • A light protection window for the valve block is also included in the scope of delivery
  • Titration impulse triggered by the motor enables the achievement of a droplet break up from 10 μL for precision determination
  • Media recirculating system for simple and rapid venting without reagent loss
  • Free rotation
  • TFT touchscreen with user-friendly menu
  • Method memory for nine different titration procedures with storage of all settings
  • USB and RS232 interface for PC connection
  • Integration in automated procedures possible

Specifications

Display Type TFT Touchscreen
Volume (Metric) Sample 50 mL
Voltage 100/240 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.