missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Orange G, 85+%, pure, high purity biological stain

Product Code. 10742081 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10742081 25 g Glass bottle 25g
10286950 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10742081 Supplier Thermo Scientific Chemicals Supplier No. 229820250
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1936-15-8 | C16H10N2Na2O7S2 | 452.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1936-15-8
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.36
MDL Number MFCD00012457
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym Acid Orange 10, 7-Hydroxy-8-(phenylazo)-1, 3-naphthalenedisulfonic acid, disodium salt, C.I. 16230
PubChem CID 131850556
IUPAC Name (8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonic acid;sodium
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O

Specifications

Dye Content 85.0% min.
Assay Percent Range 85+%
Beilstein 16, 300
Infrared Spectrum Authentic
Formula Weight 452.35
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Orange to Orange/Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Orange G
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.