missing translation for 'onlineSavingsMsg'
Learn More

Orange G, Electrophoresis Grade

Product Code. 15415059
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415059 25 g 25g
15425059 100 g 100g
15435059 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15415059 Supplier Thermo Scientific Alfa Aesar Supplier No. J62743.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Collagen stain for connective tissue | CAS: 1936-15-8 | C16H10N2Na2O7S2 | 452.36 g/mol

Collagen stain for connective tissue

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1936-15-8
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.36
MDL Number MFCD00012457
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym C.I. 16230; Acid Orange 10
PubChem CID 131850556
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O

Specifications

Beilstein 4120705
Solubility Information Soluble in water or methyl cellosolve; Slightly soluble in ethanol
IUPAC Name disodium (8Z)-7-oxo-8-(2-phenylhydrazin-1-ylidene)-7,8-dihydronaphthalene-1,3-disulfonate
Formula Weight 452.38
Grade Electrophoresis
Color Orange to Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Orange G

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.