missing translation for 'onlineSavingsMsg'
Learn More

Orange G Sodium Salt, MP Biomedicals

Product Code. 11432582 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432582 25 g 25g
11442582 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432582 Supplier MP Biomedicals Supplier No. 0215178725
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1936-15-8
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.36
MDL Number MFCD00012457
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym Acid orange 10, 7- hydroxy-8- phenylazo-1, 3- naphthalenedisulfonic acid
PubChem CID 131850556
IUPAC Name disodium (8Z)-7-oxo-8-(2-phenylhydrazin-1-ylidene)-7,8-dihydronaphthalene-1,3-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O

Specifications

Formula Weight 452.4
Quantity 25 g
Chemical Name or Material Orange G Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.