missing translation for 'onlineSavingsMsg'
Learn More

Orange G Sodium Salt, Pure, Fisher Chemical

Product Code. 10030270 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass Jar
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10030270 25 g Glass Jar 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10030270 Supplier Fisher Chemical Supplier No. O/0275/46
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 141°C
CAS 1936-15-8
Color Orange
Physical Form Solid
Quantity 25 g
Molecular Formula C16H10N2Na2O7S2
MDL Number MFCD00012457
Synonym orange g, 7-hydroxy-8-phenylazo-1,3-naphthalenedisulfonic acid, sodium e-7-hydroxy-8-phenyldiazenyl naphthalene-1,3-disulfonate
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O
IUPAC Name (8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonic acid;sodium
Molecular Weight (g/mol) 452.36
PubChem CID 131850556
Formula Weight 452.35g/mol
Packaging Glass Jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.