missing translation for 'onlineSavingsMsg'
Learn More

Orange G, Technical Grade, TRC

Product Code. 30419049
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30419049 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419049 Supplier Toronto Research Chemicals Supplier No. TRCO670600100MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acid Orange 10
CAS 1936-15-8
Molecular Formula C16 H10 N2 O7 S2 . 2 Na
Quantity 100 mg
Synonym 1,3-Naphthalenedisulfonic acid','7-hydroxy-8-(phenylazo)-','disodium salt (9CI), 2-Naphthol-6,8-disulfonic acid','1-phenylazo-','disodium salt (6CI), C.I. Acid Orange 10 (7CI), C.I. Acid Orange 10','disodium salt (8CI), 1-(Phenylazo)-2-naphthol-6,8-disulfonic acid disodium salt, 1370 Orange, Acid Crystal Orange, Acid Fast Orange EGG, Acid Fast Orange G, Acid Fast Orange GG, Acid Leather Orange G, Acid Leather Orange KG, Acid Leather Orange PGW, Acid Lightfast Orange, Acid Orange 10, Acid Orange 2G, Acid Orange 2GL, Acid Orange EGG, Acid Orange G, Acid Orange G 100, Acid Orange G Crystal, Acid Orange GG, Acid Orange Lightfast, Acidal Fast Orange, Acilan Orange GX, Amacid Crystal Orange, Amacid Light Orange G, Anadurm Orange A-G, Apocid Orange 2G, Atul Acid Crystal Orange G, Brasilan Orange 2G, Bucacid Fast Orange G, C.I. 16230, C.I. Food Orange 4, Calcocid Fast Light Orange 2G, Certicol Orange GS, Cetil Light Orange GG, Colacid Orange G, Colocid Orange 2G, Colorosacid Orange 10, Concorde Acid Orange L-GX, Crystal Orange 2G, Crystal Orange G, D and C Orange No. 3, Dinacid Crystal Orange G, Dolkwal Orange G, Duasyn Acid Orange GG, Dyacid Orange G, Dynacid Orange 2G, Egacid Orange GG, Eniacid Light Orange G, Erio Fast Orange AS, Fabracid Orange FL, Fast Acid Orange G, Fast Light Orange G, Fast Light Orange GA-CF, Fenazo Light Orange 2G, Food Orange 4, Food Orange GG, Hexacol Orange G, Hexacol Orange GG Crystals, Hidacid Fast Orange G, Hispacid Fast Orange 2G, Indacid Crystal Orange, Kemacid Crystal Orange G, Keyacid Orange 2G, Kiton Fast Orange G, Light Orange G, Multacid Orange 2G, Naphthalene Fast Orange 2G, Naphthalene Fast Orange 2GS, Naphthazine Orange 2G, Neelicol Orange G, Neklacid Fast Orange GG, Orange 2G, Orange BPC, Orange G, Orange G II, Orange GBPC, Orange GG, Pacid Orange G 10, Sandolan Fast Orange E-GL, Sandolan Orange E-GL, Solar Light Orange GX, Straight Orange G, Tertracid Light Orange G, Triacid Light Orange 2G, Vicoacid Orange IJ, Victacid Orange IJ, Vondacid Light Orange 2G, Water Orange 145924, Wool Orange 2G, Wool Orange G, Xylene Fast Orange G
InChI Formula InChI=1S/C16H12N2O7S2.2Na/c19-13-7-6-10-8-12(26(20,21)22)9-14(27(23,24)25)15(10)16(13)18-17-11-4-2-1-3-5-11;;/h1-9,19H,(H,20,21,22)(H,23,24,25);;/q;2*+1/p-2
SMILES [Na+].[Na+].Oc1ccc2cc(cc(c2c1N=Nc3ccccc3)S(=O)(=O)[O-])S(=O)(=O)[O-]
IUPAC Name disodium;7-hydroxy-8-[(E)-phenyldiazenyl]naphthalene-1,3-disulfonate
Molecular Weight (g/mol) 452.37
Formula Weight 451.9725
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.