Learn More
Cochranes Of Oxford™ Orbit Lattices Class Set
Product Code. 12869485
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Product Code. Quantity
12869485 1 Ea.
1 options
Product Code. 12869485 Supplier Cochranes Of Oxford™ Supplier No. 0048
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
Includes Diamond, graphite, the metals, sodium chloride, caesium chloride, zinc blende, wurzite, rutile, ice and layered structures
Quantity 1 Ea.
Product Type Orbit Lattices Class Set