missing translation for 'onlineSavingsMsg'
Learn More

Orcein Staining, Pure, Fisher Chemical

Product Code. 10449180 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10449180 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10449180 Supplier Fisher Chemical Supplier No. O/0300/44

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1400-62-0
Molecular Formula C28H24N2O7
MDL Number 62310
InChI Key VPEASJIRGSVXBF-UHFFFAOYSA-N
SMILES CC1=CC(=CC(=C1NC2=C(C3=C4C(=C(C(=O)C=C4OC3=CC2=O)NC5=C(C=C(C=C5C)O)O)C)C)O)O
IUPAC Name 2,8-bis(2,4-dihydroxy-6-methylanilino)-1,9-dimethyldibenzofuran-3,7-dione
Molecular Weight (g/mol) 500.507
PubChem CID 5386447
Formula Weight 500.51g/mol
Packaging Glass Jar
Color Brown/Red
Quantity 5 g
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • Orcein
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.