missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Oregon Green™ 488 Carboxylic Acid, Succinimidyl Ester, 5-isomer

Product Code. 11539186
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11539186 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11539186 Supplier Invitrogen™ Supplier No. O6147

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oregon Green™ 488 Carboxylic Acid, Succinimidyl Ester, 5-isomer

This single 5-isomer preparation of our proprietary, amine-reactive dye, Oregon Green 488 carboxylic acid, succinimidyl ester and its conjugates have green fluorescence similar to that of fluorescein but are more photostable. Additionally, Oregon Green 488 dye has a lower pKa than fluorescein (pKa = 4.7 versus 6.4 for fluorescein), making this bright dye less pH sensitive in the physiological range.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type Carboxylic Acid Succinimidyl Ester
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Oregon Green™ 488
Label Type Classic Dyes
Product Line Oregon Green

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.