missing translation for 'onlineSavingsMsg'
Learn More

Org 27569, MedChemExpress

Product Code. 16495618
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495618 10 mg 10mg
16481787 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16495618 Supplier Medchem Express Supplier No. HY13288
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Org 27569 is a potent CB1 receptor allosteric modulator, which increases agonist binding, yet blocks agonist-induced CB1 signaling.

Org 27569 is a potent CB1 receptor allosteric modulator, which increases agonist binding, yet blocks agonist-induced CB1 signaling.

Specifications

Chemical Name or Material Org 27569
CAS 868273-06-7
Color Pink
Purity Grade Notes Research
Molecular Formula C24H28ClN3O
Quantity 10 mg
Solubility Information DMSO : ≥ 52.2 mg/mL (127.33 mM)
SMILES ClC1=CC=C(C2=C1)NC(C(NCCC3=CC=C(C=C3)N4CCCCC4)=O)=C2CC
Molecular Weight (g/mol) 409.95
Formula Weight 409.95
Percent Purity 98.18%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.