missing translation for 'onlineSavingsMsg'
Learn More

3M™ Organic Vapor Monitors

Product Code. 15730018
Change view
Click to view available options
Product Type:
Organic Vapor Monitor
Organic Vapor Monitor only
Organic Vapor Monitor with Backup and Analysis
Organic Vapor Monitor with Prepaid Analysis
Unit Size:
Pack of 10
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
15730018 Organic Vapor Monitor Pack of 5
15720038 Organic Vapor Monitor with Prepaid Analysis Pack of 5
10054324 Organic Vapor Monitor only Pack of 10
15779767 Organic Vapor Monitor with Backup and Analysis Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15730018 Supplier 3M™ Supplier No. 7000144896

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lightweight, self-contained monitors record time-weighted average (TWA) vapor concentrations in factories and industrial, histology, cytology, and clinical labs

  • Provide low per-test cost. Short-term and workshift sampling can be conducted easily so you can quantify vapor exposure by employee, job category, or area.
  •  
  • Usage
  •  
    • To use, simply remove a monitor from its package and clip it to clothing near the breathing zone
    • For monitoring a specific work area, place the monitor in a location where airflow is unrestricted
    • After monitoring, seal the badge with the supplied cap; replace it in the package; record the time, date, and vapors to be analyzed; and send it to the lab for analysis†
    •  
    • Applications
    • Periodic sampling and screening of hazardous areas and jobs; indicating trouble spots; checking ventilation; and keeping accurate employee health records.
       
    • Monitor Types
      • Monitors 3500 and 3510 are designed to provide precise TWA concentration measurements for an eight-hour period. Monitors 3500 and 3520 allow you to collect data for analysis by an independent laboratory. Monitors 3510 and 3530 include prepaid analysis for up to three compounds per badge
      • Monitors 3520 and 3530 are designed with a special backup section for sampling environments that include compounds like acetone, methylene chloride, and isopropyl alcohol
         
      • Acetone, methyl ethyl ketone, and vinyl acetate may show a decreased recovery when sampled in high relative humidity. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
      • 3M Catalog# 3500, 3510, 3520, 3530

      Misuse may result in sickness or death.

    Order Info

    Data is available from 3M on sampling rate and measuring range for more than 135 vapors that these monitors can sample. Contact your Fisher Scientific Customer Service Representative at 1-800-772-6733 for a complete list of organic compounds sampled.

    Compliance

    Monitors meet or exceed OSHA accuracy requirements of ±25% for many workplace contaminants.

    The analysis service will analyze only the compounds shown in the 3M Compounds Analysis Chart. Up to three of the listed compounds can be analyzed per monitor with prepaid analysis. For analysis of unlisted compounds, contact your Fisher Customer Service Representative at 1-800-772 6733.

    TRUSTED_SUSTAINABILITY

    Specifications

    Color Black
    For Use With (Application) Periodic Sampling and Screening of hazardous areas and jobs, indicating trouble spots, Checking ventilation, and keeping accurate employee health records
    Certifications/Compliance OSHA
    Detectable Analytes Organic Vapor
    Product Type Organic Vapor Monitor
    Model 3520
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.