Learn More
Corning™ Organoid Counting Software for the Corning™ Cell Counter
Product Code. 17707024 Shop All Corning Products
Change view
Click to view available options
For Use With (Equipment):
Corning™ Cell Counter
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17707024 Corning™ Cell Counter Each
1 options
Product Code. 17707024 Supplier Corning™ Supplier No. 6749OC
Estimated Shipment: 25-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Accurate – powered by machine learning artificial intelligence.
The Organoid counting software ships with a 0.2 mm, 20 μL, counting chamber and an alphanumeric access code. This software extension enables Corning™ Cell Counter operators to capture rapid data of 3D cell cultures based on the structure’s morphology. Results are viewed within the intuitive application interface or remotely accessible by utilizing the cloud login. Unlimited storage.
  • Accurate – powered by machine learning artificial intelligence
  • Multi-informative – organoids per milliliter of solution and surface area data
  • Fast – three-second analysis utilizing cloud-based processing
  • Cost-efficient and versatile – compatible with standard 0.1 mm counting chambers and 0.2 mm counting chambers. No consumables required.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cell Counting Software
For Use With (Equipment) Corning™ Cell Counter
Includes 0.2 mm, 20 μL, counting chamber and an alphanumeric access code