missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Organoid Essentials Package, mini, PeproTech®

Product Code. 30245109 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30245109 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30245109 Supplier Gibco™ Supplier No. OREM
  • ONLINE EXCLUSIVE PRICE
    724.00 €
    521.28 € / Pack of 1
    Save 202.72 €
    28% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Organoid Essentials Package, mini, contains the key components required for developing organoid models in cell culture.

The Organoid Essentials Package, mini, contains the key components required for the development of organoid models in cell culture.

The package contains the following five vials:

  • 10 μg Mouse Wnt-3a (Cat. No. 315-20-10UG)
  • 20 μg Human Noggin (Cat. No. 120-10C-20UG)
  • 20 μg Human R-Spondin 1 (Cat. No. 120-38-20UG)
  • 25 μg Human FGF-10 (Cat. No. 100-26-25UG)
  • 100 μg Animal-Free Human EGF (Cat. No. AF-100-15-100UG)

FGF-10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. FGF-10 plays a crucial role in epithelial-mesenchymal interactions, which are essential for the development of various organs.

EGF (epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli or to EGF binding to the receptor. EGF promotes the rapid proliferation of epithelial progenitor cells, which is essential for the initial growth and expansion of organoids.

R-Spondin-1 is a member of the R-spondin family and encodes a secreted activator protein with two cysteine-rich, furin-like domains and one thrombospondin type 1 domain.

Wnt3a, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. Wnt-3A supports the growth and morphogenesis of organoids by encouraging the proliferation and differentiation of progenitor cells into various cell types, contributing to the development of complex tissue structures.

TRUSTED_SUSTAINABILITY

Specifications

Format Lyophilized
Product Type Stem Cell Expansion Cytokine Package
Quantity 1 Ea.
Species Human, Murine

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.