missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Original Perfusor™ Syringe

Product Code. 10294262
Change view
Click to view available options
Volume (Metric):
20 mL
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10294262 20 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10294262 Supplier B Braun™ Supplier No. 8728615
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Made of polypropylene. B Braun™ Original Perfusor™ Syringe complement the excellent conveying characteristics of the Perfusor™ syringe pump.

  • For use with Perfusor™syringe pumps
  • Made of polypropylene
  • Smooth piston plunger made of synthetic material, with double sealing ring
  • Minimal residual volume
  • Easy-to-read, smudge-proof graduation
  • Latex-Free / PVC-free
  • Luer lock adapter
  • Individually sterile packed

Specifications

Certifications/Compliance ISO 10993 or DIN EN 30993 and EN ISO 7886-2
Material Polypropylene
Color Clear
Latex Free Yes
Graduations 1.0 mL
Volume (Metric) 20 mL
For Use With (Equipment) Perfusor Syringe Pumps
Graduated Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.