missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Orion™ ROSS Ultra™ Thin Stem Refillable Epoxy Bodied Combination pH Electrode
Description
Thermo Scientific™ Orion™ ROSS Ultra™ Epoxy Semi-Micro Electrodes provide optimum accuracy and unmatched response in samples of varying temperatures. They are available in epoxy bodies for ruggedness and best suited for measuring samples in test tubes and small containers. Orion™ ROSS Ultra™ Epoxy Semi-Micro Electrodes comes with two year warranty period.
Orion™ ROSS Ultra™ Electrodes offer the best stability and drift free measurements of all our ROSS electrodes while still providing the rapid accuracy and temperature response synonymous with this reference system These are compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.
Solutions required
Electrode fill solution
810007 ROSS pH electrode fill solution, 5 x 60 mL bottles
Electrode storage solution
810001 ROSS pH electrode storage solution, 475 mL bottle
pH buffers
910168 pH 1.68 buffer, NIST traceable, 475 mL
910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112 pH 12.46 buffer, NIST traceable, 475 mL
810199 All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH
electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
storage bottle
electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
storage bottle
pH cleaning solution kit
900020 Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with
810007 ROSS pH electrode fill solution 60 mL 1 bottle
Warranty period
24 months from date of purchase
Specifications
Specifications
| Description | Epoxy body, semi-micro tip combination electrode; 165 x 6mm |
| Connector Types | BNC (Waterproof) |
| Junction | Glass Fiber |
| Material | Epoxy Body |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 100°C |
| Type | Electrode |
| Filling Solution Type | 3M KCl |
| Internal Reference | ROSS |
| Precision | 0.01 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?