missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ROSS Ultra™ Flat Surface Refillable Epoxy Bodied Combination pH Electrode

Product Code. 11732767 Shop All Thermo Scientific Products
Change view
Click to view available options
Connector Types:
BNC (Waterproof)
Unit Size:
Each
Product Code. Connector Types unitSize
11732767 BNC (Waterproof) Each
1 options
This item is not returnable. View return policy

Product Code. 11732767

Brand: Thermo Scientific™ 8135BNUWP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Measure flat, soft, moist, semi-solid or small volume samples with epoxy body flat surface pH electrodes.

Thermo Scientific™ Orion™ ROSS Ultra Flat Surface Electrodes have a uniquely designed, flat sensing bulb that allow for reliable pH measurement on samples such as paper, textiles, skin, meat, cheese, or in agar gel plates.
 

Orion™ ROSS Ultra Electrodes offer the best stability and drift free measurements of all our ROSS electrodes while still providing the rapid accuracy and temperature response synonomous with this reference system These are compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.
 
Solutions required 
Electrode fill solution
810007     ROSS pH electrode fill solution, 5 x 60 mL bottles
 
Electrode storage solution
810001     ROSS pH electrode storage solution, 475 mL bottle
 
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
810199     All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH
                 electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
                 storage bottle
 
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
 
Items included with 
810007     ROSS pH electrode fill solution 60 mL 1 bottle
 
Warranty period
24 months from date of purchase
TRUSTED_SUSTAINABILITY

Specifications

Description Ultra pH Electrode
Connector Types BNC (Waterproof)
Junction Glass Fiber
Material Epoxy Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type pH Electrode
Electrode Flat surface tip epoxy body combination electrode; 20 x 12mm
Filling Solution Type 3M KCl
Internal Reference ROSS
Precision 0.01
Temperature (English) Operating 32°F to 212°F
Warranty 2 Years
Length (English) Cable 39 in.
Length (Metric) 1 m
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.