missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Carbon Dioxide Combination Ion Selective Electrode

Product Code. 11996069
Change view
Click to view available options
Connector Types:
BNC (Waterproof)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
11996069 BNC (Waterproof) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11996069 Supplier Thermo Scientific™ Supplier No. 9502BNWP
  • ONLINE EXCLUSIVE PRICE
    979.88 €
    781.20 € / Pack of 1
    Save 198.68 €
    20% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Measure carbon dioxide, carbonate and bicarbonate in aqueous solutions quickly, accurately and economically.

Measure carbon dioxide, carbonate and bicarbonate in aqueous solutions quickly and accurately with the Thermo Scientific™ Orion™ 9502BNWP Carbon Dioxide Electrode. 

Features:

  • It features a gas-permeable membrane technology for use with loose membranes and O-rings.

Solutions required:

  • Standard
    • 950206 0.1 M NaHCO3 Carbon Dioxide standard, 475 mL
    • 950207 1000 ppm as CaCO3 Carbon Dioxide standard, 475 mL
  • Carbon dioxide buffer solution
    • 950210 Carbon Dioxide Ionic Strength Adjuster (ISA), 475 mL
  • Electrode fill solution
    • 950202 Carbon Dioxide electrode fill solution, 60 mL
  • Storage solution
    • 941706 0.1 M NaCl Chloride standard, 475 mL

Items included with:

  • 950202 Carbon Dioxide electrode fill solution, 60 mL 1 bottle
  • 950204 4 membranes with 1 O-ring

Warranty period:

  • 12 months from date of purchase
TRUSTED_SUSTAINABILITY

Specifications

Connector Types BNC (Waterproof)
Description Carbon Dioxide Electrode
Junction Annular PTFE
Material Epoxy
For Use With (Equipment) ISE Meter with a BNC Connection
Temperature (Metric) Operating 0°C to 50°C
Type Carbon Dioxide Electrode
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.