missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AQUAfast™ AQ4500 Turbidity Meter Accessories

Product Code. 11757717
Change view
Click to view available options
Type:
Cable
Formazin Turbidity Standard
Replacement Turbidity Vial
Silicone Oil and Cloth Kit
Standards
Unit Size:
475mL
Each
Pack of 12
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11757717 Formazin Turbidity Standard 475mL
10494362 Standards Each
11767717 Silicone Oil and Cloth Kit Each
11722178 Replacement Turbidity Vial Pack of 12
10672853 Cable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11757717 Supplier Thermo Scientific™ Supplier No. AC45FZ
  • ONLINE EXCLUSIVE PRICE
    92.65 €
    77.40 € / 475mL
    Save 15.25 €
    16% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Ensure your turbidity meter has the accessories it needs, including turbidity vials, primary turbidity standards, Formazin turbidity standards, and more.

Ensure your Thermo Scientific™ Orion™ AQUAfast™ AQ4500 Turbidity Meter has the accessories it needs with our selection of Orion™ AQUAfast™ AQ4500 Turbidity Meter Accessories that include turbidity vials, primary turbidity standards, Formazin turbidity standards, and more. Our Orion AQUAfast AQ4500 turbidity meter has advanced features, including dual light sources and drinking water / wastewater methods approved for U.S. EPA regulatory reporting. Keep pace with changing regulatory protocols; use the White Light LED to comply with EPA 180.1 requirements or Infrared (IR) LED to comply with ISO 7027 requirements. Select from measure modes including EPA 180.1, ISO Nephelometric, ISO Absorbance, % transmittance, EBC and ASBC. This IP67 waterproof meter records 100 data points and offers an average battery life of 2500 hours intermittent use.

Log data into internal memory, or order AQ4CBL cable to send data to printer or download to a computer for easy time and date stamped results storage.
  • U.S. EPA approval for Drinking Water Reporting per 40 CFR Part 141, vol. 74, no. 147, Monday, August 3, 2009. Rules and regulations pg. 38351-38356
  • Dual source to comply with EPA 180.1 and ISO 7027
  • Nephelometric and ratiometric measurements with autoranging
  • IP67 waterproof portable turbidity meter
  • Readings in the range of 0 to 1000 NTU with a choice of units: NT, FTU, FNU, ASBC or EBC

Includes:

  • AQ4500 meter
  • Set of 5 USEPA approved primary calibration standards
  • Seven glass vials
  • User guide
  • Carrying case
TRUSTED_SUSTAINABILITY

Specifications

Type Formazin Turbidity Standard
For Use With (Equipment) AQ4500 Turbidimeter
Description 4000NTU, 475mL
EPA Approved Methods US EPA approval for Drinking Water Reporting per 40 CFR Part 141, vol. 74, no. 147, Monday, August 3, 2009. Rules and regulations pg 38351-38356.EPA 180.1 and ISO 7027
compliance-icons
Product Identifier
  • Formazin Standard, 4000 NTU
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see supplemental first aid instructions on this label).
Supplemental information
  • MIXTURE LIST-Contain: Hexamethylenetetramine

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.