Learn More
Thermo Scientific™ Orion™ AQUAfast™ Free & Total Chlorine Bulk DPD Powder Reagents
Description
Measure residual chlorine in water samples using Thermo Scientific™ Orion™ AQUAfast Free and Total Chlorine Bulk DPD Powder Dispensers and Vials with Orion AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers. The powder chemistries dissolve quickly in the solution and are easy to mix. Each vial contains 250 counts of DPD reagent and is available for free chlorine or total chlorine analysis in the lab or field.
- Measure 0.02 to 2.00 mg/L (ppm) free chlorine or total chlorine
- Bulk powder dispensers are designed for high sample throughput – reduces the cost per test and minimizes waste compared with individually packaged reagents
- Equivalent to other widely used and recognized chemistries including Orion AQUAfast AC4P71 free chlorine and Orion AQUAfast AC4P72 total chlorine powder packs
- Compatible with Orion AQUAfast AQ4000, AQ3700 and AQ3170 colorimeters
- Compatible with Orion AquaMate AQ8000 and AQ7000 spectrophotometers
Spécification
Spécification
| For Use With (Equipment) | AQUAfast AQ250T total chlorine DPD bulk powder dispenser |
| Includes | DPD Powder Refill for AQ250T |
| Quantity | 2 x 250 count |
| Range | 0.02 to 2 mg/L Cl2 |
| Reagent Form | Powder |
| Reagents | Chlorine |
| Test Method | DPD |
Sécurité et traitement
- AQUAfast total chlorine powder pack
- Danger||Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- H315-Causes skin irritation.
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P391-Collect spillage.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P403-Store in a well-ventilated place.
- P270-Do not eat, drink or smoke when using this product.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
- MIXTURE LIST-Contain: sodium oxalate, N,N-diethylbenzene-1,4-diammonium sulphate
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.