Learn More
Thermo Scientific™ Orion™ AQUAfast™ Free & Total Chlorine Bulk DPD Powder Reagents
Product Code. 15954002
Change view
Click to view available options
Includes:
AQ250F Dispenser, 1 vial
AQ250T Dispenser, 1 vial
DPD Powder Refill for AQ250F
DPD Powder Refill for AQ250T
Quantity:
2 x 250 count
250 count
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Quantity Unit Size
15954002 AQ250F Dispenser, 1 vial 250 count Each
15914002 DPD Powder Refill for AQ250F 250 count Each
15924002 DPD Powder Refill for AQ250F 2 x 250 count Each
15964002 AQ250T Dispenser, 1 vial 250 count Each
15934002 DPD Powder Refill for AQ250T 250 count Each
15944002 DPD Powder Refill for AQ250T 2 x 250 count Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15954002 Supplier Thermo Scientific™ Supplier No. AQ250F
  • ONLINE EXCLUSIVE PRICE
    80.75 €
    66.78 € / Pack of 1
    Save 13.97 €
    17% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Thermo Scientific™ Orion™ AQUAfast Free and Total Chlorine Bulk DPD Powder Dispensers and Vials are designed for high volume testing of residual chlorine in water samples.
Measure residual chlorine in water samples using Thermo Scientific™ Orion™ AQUAfast Free and Total Chlorine Bulk DPD Powder Dispensers and Vials with Orion AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers. The powder chemistries dissolve quickly in the solution and are easy to mix. Each vial contains 250 counts of DPD reagent and is available for free chlorine or total chlorine analysis in the lab or field.
  • Measure 0.02 to 2.00 mg/L (ppm) free chlorine or total chlorine
  • Bulk powder dispensers are designed for high sample throughput – reduces the cost per test and minimizes waste compared with individually packaged reagents 
  • Equivalent to other widely used and recognized chemistries including Orion AQUAfast AC4P71 free chlorine and Orion AQUAfast AC4P72 total chlorine powder packs
  • Compatible with Orion AQUAfast AQ4000, AQ3700 and AQ3170 colorimeters
  • Compatible with Orion AquaMate AQ8000 and AQ7000 spectrophotometers
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 colorimeter, Orion AQUAfast AQ3700 colorimeter, Orion AQUAfast AQ3170 colorimeter, Orion AQUAfast AQ3070 (obsolete)
Includes AQ250F Dispenser, 1 vial
Quantity 250 count
Range 0.02 to 2 mg/L Cl2
Reagent Form Powder
Reagents Chlorine
Test Method DPD
compliance-icons compliance-icons compliance-icons
Product Identifier
  • AQUAfast total chlorine powder pack
Signal Word
  • Danger||Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P391-Collect spillage.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P403-Store in a well-ventilated place.
  • P270-Do not eat, drink or smoke when using this product.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contain: sodium oxalate, N,N-diethylbenzene-1,4-diammonium sulphate