Learn More
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents
Beskrivning
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
- No. of test: 30
- Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
- Thermo Scientific™ Auto-ID™ eliminates measurement errors
Specifikationer
Specifikationer
| Type | Chlorine |
| Test Reagent Formats | Ampoule |
| For Use With (Equipment) | Orion AQUAfast AQ4000 Colorimeter |
| Description | AQUAfast chlorine (free & total) Auto-ID ampoules, 30 pack |
| Includes | 30 Auto-ID Ampoules |
| Method | DPD |
| For Use With (Application) | Colorimetric water and wastewater testing equipment |
| Recommended Accessories | AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER) |
| Range | 0.4 to 5 mg/L Cl2 |
| EPA Approved Methods | Drinking Water: Standard Methods 4500-Cl G / ATP EPA-approved Orion Method AC4070; Wastewater: Standard Methods 4500-Cl G-2000 / ATP EPA-approved Orion Method AC4070 (total) |
| Visa mer |
Säkerhet och hantering
- Chlorine (Free & Total)
- Warning
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P370+P378-In case of fire: Use materials appropriate for surrounding fire to extinguish.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : acetone
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.