Learn More
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents
Product Code. 11792188
Change view
Click to view available options
Type:
Aluminum
Ammonia
Chloride
Chlorine
Chlorine Dioxide
Copper
Iron
Manganese
Nitrate
Nitrite
Ozone
Phosphate
Silica
Sulfate
Sulfide
Zinc
Range:
0.04 to 0.25 mg/L Al
0.08 to 1 mg/L N
0.2 to 1.5 mg/L N
0.2 to 3 mg/L N
0.2 to 3 mg/L S
0.2 to 5 mg/L O3
0.2 to 6 mg/L Fe
0.3 to 3 mg/L Zn
0.3 to 8 mg/L PO4
0.4 to 5 mg/L Cl2
0.5 to 10 mg/L SiO2
0.5 to 12 mg/L Cu
0.5 to 7 mg/L N
0.8 to 11 mg/L ClO2
1.0 to 7.5 mg/L N
1.5 to 14 mg/L N
10 to 100 mg/L SO4
2 to 30 mg/L Mn
2.5 to 40 mg/L Cl-
5 to 50 mg/L NO3
Method:
Bariumsulfate-Turbidity
Bathocuproine
Cadmium Reduction
DPD
Diazotization (Azo)
Direct Nesslerization
Eriochrome Cyanine R
Ferric Thiocyanide
Heteropoly Blue
Methylene Blue
Periodate
Phenanthroline
Salicylate
Stannous Chloride
Zincon
Unit Size:
30 tests
Pack of 30
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Range Method unitSize
11792188 Nitrate 0.2 to 1.5 mg/L N Cadmium Reduction 30 tests
11702198 Nitrate 1.0 to 7.5 mg/L N Cadmium Reduction 30 tests
11722198 Nitrate 5 to 50 mg/L NO3 Cadmium Reduction 30 tests
11752198 Ammonia 0.2 to 3 mg/L N Salicylate 30 tests
11762198 Ammonia 1.5 to 14 mg/L N Direct Nesslerization 30 tests
11772198 Ammonia 0.5 to 7 mg/L N Direct Nesslerization Pack of 30
11782198 Sulfide 0.2 to 3 mg/L S Methylene Blue 30 tests
11792198 Chloride 2.5 to 40 mg/L Cl- Ferric Thiocyanide 30 tests
11702208 Aluminum 0.04 to 0.25 mg/L Al Eriochrome Cyanine R 30 tests
11712208 Copper 0.5 to 12 mg/L Cu Bathocuproine 30 tests
11722208 Nitrite 0.08 to 1 mg/L N Diazotization (Azo) 30 tests
11732208 Ozone 0.2 to 5 mg/L O3 DPD 30 tests
11742208 Manganese 2 to 30 mg/L Mn Periodate 30 tests
11752208 Silica 0.5 to 10 mg/L SiO2 Heteropoly Blue 30 tests
11762208 Zinc 0.3 to 3 mg/L Zn Zincon 30 tests
11772208 Chlorine 0.4 to 5 mg/L Cl2 DPD 30 tests
11782208 Iron 0.2 to 6 mg/L Fe Phenanthroline 30 tests
11792208 Sulfate 10 to 100 mg/L SO4 Bariumsulfate-Turbidity 30 tests
11702218 Phosphate 0.3 to 8 mg/L PO4 Stannous Chloride 30 tests
11712218 Chlorine Dioxide 0.8 to 11 mg/L ClO2 DPD 30 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Product Code. 11792188 Supplier Thermo Scientific™ Supplier No. AC4004
  • ONLINE EXCLUSIVE PRICE
    142.00 €
    135.00 € / 30 tests
    Save 7.00 €
    4% Off
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
Minimize the risks of improper measurement and contamination with these chemistry reagents, which are contained in self-filling vials.
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
  • No. of test: 30
  • Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
  • Thermo Scientific™ Auto-ID™ eliminates measurement errors
TRUSTED_SUSTAINABILITY
Specifications
Type Nitrate
Test Reagent Formats Ampoule
For Use With (Equipment) Orion AQUAfast AQ4000 Colorimeter
Description AQUAfast nitrate LR Auto-ID ampoules, 30 pack
Includes 30 Auto-ID Ampoules
Method Cadmium Reduction
For Use With (Application) Colorimetric water and wastewater testing equipment
Recommended Accessories AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER)
Range 0.2 to 1.5 mg/L N
EPA Approved Methods Drinking Water: ASTM D3867-90 (B) / Standard Methods 4500-NO3- E; Wastewater: ASTM D3867-04 (B) / Standard Methods 4500-NO3- E
Quantity 30 Ampoules
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.