missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AQUAfast™ Reaction/Digestion Tubes and Liquid Reagents

Product Code. 15924012
Change view
Click to view available options
Type:
Ammonia
COD
COD High Range
COD Low Range
COD Mid Range
Fluoride
Manganese
Nitrate
Nitrogen, Total
Phosphate
pH
Range:
0 to 150 mg/L O2
0 to 1500 mg/L O2
0 to 15000 mg/L O2
0.01 to 0.7 mg/L Mn
0.02 to 1.1 mg/L P
0.02 to 1.6 mg/L P
0.02 to 2.5 mg/L N
0.05 to 2 mg/L F
0.06 to 5 mg/L PO4
0.5 to 25 mg/L N
1 to 30 mg/L N
1 to 50 mg/L N
5 to 150 mg/L N
6.5 to 8.4 pH
Unit Size:
25 tests
Each
Pack of 100
Pack of 2
Pack of 25
Pack of 50
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Range unitSize
15924012 COD Mid Range 0 to 1500 mg/L O2 Each
11742218 Fluoride 0.05 to 2 mg/L F Pack of 50
11757757 pH 6.5 to 8.4 pH Pack of 2
15169192 Manganese 0.01 to 0.7 mg/L Mn Pack of 100
15189192 Nitrogen, Total 0.5 to 25 mg/L N Pack of 50
15199192 Nitrogen, Total 5 to 150 mg/L N Pack of 50
15109202 Phosphate 0.02 to 1.1 mg/L P Pack of 50
15119202 Phosphate 0.02 to 1.6 mg/L P Pack of 50
15179192 Nitrate 1 to 30 mg/L N Pack of 50
15129192 Ammonia 1 to 50 mg/L N Pack of 50
15199182 Ammonia 0.02 to 2.5 mg/L N Pack of 50
15129202 Phosphate 0.06 to 5 mg/L PO4 Pack of 50
11722188 COD 0 to 1500 mg/L O2 25 tests
11742188 COD 0 to 15000 mg/L O2 Pack of 25
11762188 COD 0 to 150 mg/L O2 25 tests
15904012 COD High Range 0 to 15000 mg/L O2 Each
15914012 COD Low Range 0 to 150 mg/L O2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Product Code. 15924012 Supplier Thermo Scientific™ Supplier No. CODM150
Wastewater Testing Savings
Use promo code 28023
View all promotions
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Perform colorimetric testing including fluoride, COD, total nitrogen and total phosphate using the Thermo Scientific™ Orion™ AQUAfast Liquid Reagents, Digestion and Reaction Tubes.

Use Thermo Scientific™ Orion™ AQUAfast Digestion Tubes, Reaction Tubes and Liquid Reagents with Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers. Digestion and reaction tubes contain the precise amount of reagent, are made of special optical glass and fit the Orion™ AQUAfast COD165 thermoreactor. Digestion tubes measure COD, total nitrogen and total phosphate. Reaction tubes measure ammonia, nitrate and phosphate. Liquid reagents mix quickly and easily in solution and measure fluoride (SPADN), hydrazine and pH.

Changes in nutrient levels can alter the delicate balance in ecosystems and contaminate drinking water. Tube tests make it easy to determine concentrations of species in samples.

 

TRUSTED_SUSTAINABILITY

Specifications

Description AQUAfast COD 0-1500 ppm digestion tubes
For Use With (Equipment) Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast
Includes 150 Mid Range COD Digestion Tubes
Quantity 150 Tubes
Range 0 to 1500 mg/L O2
Test Method Dichromate/H2SO4
Type COD Mid Range
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • COD medium range (0-1,500 mg/l)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 A
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: sulphuric acid, mercury sulphate, disilver sulphate, potassium dichromate

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.