missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ AquaMate 8000 UV-Vis Spectrophotometer

Product Code. 14845061
Change view
Click to view available options
Type:
AquaMate 8000 Spectrophotometer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
14845061 AquaMate 8000 Spectrophotometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 14845061 Supplier Thermo Scientific™ Supplier No. AQ8000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Equip your lab with the accurate, easy to use Thermo Scientific™ Orion™ AquaMate 8000 UV-Vis Spectrophotometer to ensure the ideal instrument for water analysis and beyond

Equip your lab with the accurate, easy to use Thermo Scientific™ Orion™ AquaMate 8000 UV-Vis Spectrophotometer to ensure you have the ideal instrument for water analysis and beyond. This spectrophotometer offers 260 preprogrammed methods for ion concentration from absorbance plus wavelength scanning function and standard curve function for creating custom method using so virtually any brand of colorimetric reagent can be used. Utilize 190-1100 nm selectable wavelengths, 1.8 nm spectral bandwidth and vial flexibility with the included 3-position carousel for holding round and rectangular vials.

Pre-programmed Methods:

  • More than 260 pre-programmed methods offer easy and convenient measurements using Orion™ AQUAfast™, Merck™ or CHEMetrics™ chemistries
  • Pre-programmed methods can also be adapted for new chemistries or operators can create their own custom methods and calibration curves
  • Instruments allow a one-point adjustment on any preprogrammed method to correct for variations in batch reagent chemistries

Versatility:

  • Wide variety of vials sizes can be used, including 10mm square vials, 20 and 50mm rectangular vials and an adjustable round vial holder that accommodates 13, 16, and 24mm round vials
  • Three USB connections provide the options to store methods and data to a USB memory stick, transfer data to a computer, and print hard copy data to an external printer
  • System is compatible with ink jet and laser printers with HP PCL 5 or greater

Verification:

  • Performance verification tests ensure wavelength accuracy and instrument functionality
  • Each verification report gives the time, date and instrument serial number in accordance with GLP and GMP
  • Built-in wavelength accuracy test is compatible with either the internal lamp or external calibrated standards
  • Built-in test methods for stray light, noise and resolution are available to validate instrument performance further

Includes:

  • Methods on USB memory stick
  • Three-position cell holder
  • User documentation
  • Dust cover
  • USB cable
  • 110V, 220V and 240V power cords
TRUSTED_SUSTAINABILITY

Specifications

Compatibility Compatible with ink jet and laser printers with HP PCL 5 or greater
Connections USB type A port for USB stick (front panel)
USB type B port for computer (rear panel)
USB type A port for printer (rear panel)
Detector Type Dual Silicon Photodiodes
Display Graphical with LCD backlight, 9.7 x 7.1 cm (3.8 x 2.8 in.)
Spectral Bandwidth 1.8 nm
Type AquaMate 8000 Spectrophotometer
Voltage 100/240 V
Wavelength Range 190 to 1100 nm
Drift <0.0005 A/hr.
Electrical Requirements 100/240 V, 50/60 Hz
For Use With (Application) Sample Concentration
Height (English) 9.8 in.
Height (Metric) 25 cm
Hertz 50/60 Hz
Includes Methods on USB memory stick; 6-position cell holder; 3-position cell holder; 1-position cell holder; User documentation; Dust cover; USB cable; 110V and 220V power cords
Keypad Sealed membrane with tactile response keys
Lamp Life 5 years
Noise <0.00050 at 1.0 A; <0.00080 at 2.0 A RMS at 260nm
Optical Design Double beam—internal reference detector
Photometric Linearity Up to 3.5A at 260nm
Photometric Range -0.5 to 5.0 A ; -1.5 to 125 %T; ±9999 C
Printer Included External Printer: Ink Jet, Laser Printer with HP PCL 5 or Greater
Scan Speed 10 to 4200 nm/min.
Stray Light <0.08 %T at 220 nm and 340 nm; <0.1 %T at 340 nm and 400 nm; <1.0 %T at 198 nm
Wavelength Accuracy ±1.0 nm
Wavelength Data Interval 0.2; 0.5; 1.0; 2.0; 3.0; 5.0 nm
Wavelength Repeatability ±0.5nm
Wavelength Slew Speed 11,000 nm/min.
Weight (English) 19 lb.
Weight (Metric) 8.6 kg
Width (English) 11.8 in.
Width (Metric) 30 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.