Learn More
Thermo Scientific™ Orion™ Calibration Sleeves for Dissolved Oxygen Probes
Product Code. 12983247
Change view
Click to view available options
Type:
Calibration Sleeve
Calibration Sleeve with Stand
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
12983247 Calibration Sleeve Each
11722607 Calibration Sleeve with Stand Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12983247 Supplier Thermo Scientific™ Supplier No. 080017
  • ONLINE EXCLUSIVE PRICE
    174.87 €
    136.80 € / Pack of 1
    Save 38.07 €
    21% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Calibrate and store probes with Thermo Scientific™ Orion™ Calibration Sleeves for Dissolved Oxygen Probes.
  • Provide optimum stable environment for calibration.
Warranty: Out of Box
TRUSTED_SUSTAINABILITY
Specifications
Description Calibration Sleeve
For Use With (Equipment) DO Probe
Type Calibration Sleeve