Learn More
Thermo Scientific™ Orion™ Cyanide Electrodes
Measure cyanide ions in aqueous solutions quickly, simply, accurately and economically with these cyanide electrodes.
636.35 € - 1010.00 €
Specifications
Junction | Free-flowing liquid junction assures stable, drift-free potentials |
---|---|
For Use With (Equipment) | ISE Meters |
Product Code | Brand | Connector Types | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connector Types | Price | Quantity & Availability | |||||
10039150
|
Thermo Scientific™
9406BN |
BNC |
1010.00 €
Each |
In Stock
Log in to see stock. |
|||||
10549131
|
Thermo Scientific™
9606BNWP |
Waterproof BNC |
861.00 €
Each |
Estimated Shipment: 04-07-2024
Log in to see stock. |
|||||
10232591
|
Thermo Scientific™
9406SC |
Screw Cap |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Measure cyanide ions in aqueous solutions quickly, simply and economically with Thermo Scientific™ Orion™ Cyanide Electrodes. No separate reference electrode is needed with this electrode, making it convenient to use with small sample volumes.
No separate reference electrode is needed with this electrode, making it convenient to use with small sample volumes.- Calibration Standards Catalog number: Consult instruction manual
- Required ISA Catalog number: 951011
Warranty: 12 months from date of purchase by Buyer or 18 months from date of shipment from Seller, whichever is earlier.
Specifications
Free-flowing liquid junction assures stable, drift-free potentials | |
ISE Meters |