missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Combination pH Electrodes

Product Code. 11547054
Change view
Click to view available options
Filling Solution Type:
Gel-filled
Liquid-filled
Junction:
Double
Single
Unit Size:
Each
Product Code. Filling Solution Type Junction unitSize
11547054 Gel-filled Double Each
10359834 Liquid-filled Double Each
11567054 Gel-filled Single Each
10709185 Liquid-filled Single Each
4 options
This item is not returnable. View return policy

Product Code. 11547054

Brand: Thermo Scientific™ GD9106BNWP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Environmentally friendly lab pH electrodes to meet RoHS requirements by containing no lead, mercury or other hazardous substances — making disposal of old electrodes easy and hassle free.

Thermo Scientific™ Orion™ Green Electrodes are the first lab electrode to contain no lead, mercury or other hazardous substances and are completely ROHS compliant. GD9156BNWP is a refillable electrode that provides reliable and accurate measurements of routine samples but are best suited for treated effluent, distilled water and wastewater. GD9106BNWP and GS9106BNWP contain a low-maintenance gel that is best suited for field or plant use and educational institutes, where minimal maintenance is required.
 

  • No disposal issues, no disposal cost
  • Choice of single-junction models for clean-water measurements, or double-junction models for dirty-water testing
  • Choice of refillable or low-maintenance gel-filled models
  • Duarable epoxy body
  • pH Range: 0 to 14
  • Dimensions: 120 × 12mm, cap dia. 16mm
  • Waterproof BNC connector, 1m cable; fit any meter with BNC input
Warranty:
  • 12 months from date of purchase (GD9156BNWP)
  • 6 months from date of purchase (GS9106BNWP and GD9106BNWP)

Warranty

  • 12 months from date of purchase (GD9156BNWP)
  • 6 months from date of purchase (GS9106BNWP and GD9106BNWP).

TRUSTED_SUSTAINABILITY

Specifications

Description Low-maintenance gel-filled
Connector Types BNC (Waterproof), BNC-to-USS adapter is required with this product as a onetime purchase. Part Number: 13-641-890
Junction Double
Material Epoxy Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 90°C
Type Green Combination pH Electrode, gel-filled
Filling Solution Type Gel-filled
For Use With (Equipment) Orion pH Meter
Length (English) Cable 39.3 in.
Length (Metric) Cable 1 m
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.