missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ISE Filling Solutions

Product Code. 12024502
Change view
Click to view available options
Quantity:
60 mL
5 x 60 mL
For Use With (Equipment):
Ammonia ISE
Ammonium ISE
Carbon Dioxide ISE
Nitrate ISE
Potassium ISE
Sodium ISE
Sodium ISE (Low Sodium)
Unit Size:
60mL
Each
Pack of 5
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity For Use With (Equipment) Unit Size
12024502 60 mL Ammonia ISE 60mL
12084502 5 x 60 mL Sodium ISE Pack of 5
12048882 5 x 60 mL Sodium ISE (Low Sodium) Pack of 5
11762787 5 x 60 mL Ammonium ISE Pack of 5
12074502 5 x 60 mL Nitrate ISE Pack of 5
11737797 5 x 60 mL Potassium ISE Pack of 5
11717677 60 mL Carbon Dioxide ISE 60mL
10675261 60 mL Ammonia ISE Each
12871521 60 mL Ammonia ISE Each
10368883 60 mL Ammonia ISE 60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 12024502 Proveedor Thermo Scientific™ N.º de proveedor 951202
  • ONLINE EXCLUSIVE PRICE
    89.25 €
    76.68 € / 60mL
    Save 12.57 €
    14% Off
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Enhance the performance and extend the life of your ion selective electrodes with these ISE filling solutions.

Enhance the performance and extend the life of your ion selective electrodes with Thermo Scientific™ Orion™ ISE Filling Solutions. These solutions are specially formulated to work with the recommended ion selective electrodes and prevent electrode interferences from filling solution.

Features:

  • Filling/maintenance solutions are for use with Thermo Scientific™ Orion™ Ion Selective Electrodes (ISE)
  • Optimum Result™ ISE filling solutions are specially designed for the Ionplus™ series ion selective electrodes

Ordering Information:

  • Packaged as set of five 2 oz. (5 x 60mL) bottles, except Cat. No. 951209, 951202 and 950202 packaged as one 2 oz. (60mL) bottle and Cat. No. 954602 packaged as one 1.6 oz. (50mL) bottle

Note:

The following part number has been changed due to a recent system upgrade:

Old Part Number: 900018

New Part Number: 900018-WA

Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.

TRUSTED_SUSTAINABILITY

Especificaciones

Description Fill Solution For Ammonia ISE
For Use With (Equipment) Ammonia ISE
Type ISE Filling Solution
Quantity 60 mL
compliance-icons compliance-icons
Nombre del producto
  • Ammonia Electrode Filling Solution
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P202-Do not handle until all safety precautions have been read and understood.
  • P273-Avoid release to the environment.
  • P321-Specific treatment (see supplemental first aid instructions on this label).
Otros peligros
  • MIXTURE LIST-Contain: Ammonium Chloride, Silver Nitrate

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.