missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents

Product Code. 11777527
Change view
Click to view available options
Type:
Acid Buffer
Acid Reagent
Alkaline Reagent
CISA
ISA Solution
Iodide Reagent
Ionic Strength Adjustor Buffer
Nitrate Interference Suppressor
Storage Solution
Sulfide Anti-Oxidant Buffer
TISAB II
TISAB III
pH Adjusting ISA
pH Adjusting Low Level ISA
For Use With (Equipment):
Ammonia Electrode
Bromide Electrode, Cadmium Electrode, Chloride Electrode, Cupric Electrode, Iodide Electrode, Sufide Electrode, Thiocyanate Electrode
Calcium Electrode
Carbon Dioxide Electrode
Chloride Electrode
Chlorine Electrode
Cyanide Electrode
Fluoride Electrode
Fluoroborate Electrode, Nitrate Electrode, Perchlorate Electrode
Nitrate Electrode
Nitrogen Oxide Electrode
Potassium Electrode
Sodium Electrode
Sodium ISE
Sulfide Electrode
Quantity:
475 mL
5 x 50 mL
1 Gal.
4 x 475 mL
2 x 1 L
4 x 1 Gal.
Unit Size:
3.8 L
475mL
Each
Pack of 2
Pack of 4
Pack of 5
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Quantity Unit Size
11777527 ISA Solution Fluoroborate Electrode, Nitrate Electrode, Perchlorate Electrode 475 mL 475mL
11722787 Storage Solution Sodium Electrode 475 mL Each
12049967 Ionic Strength Adjustor Buffer Sodium ISE 475 mL 475mL
11767527 Nitrate Interference Suppressor Nitrate Electrode 475 mL 475mL
11717537 ISA Solution Potassium Electrode 475 mL 475mL
11747537 ISA Solution Calcium Electrode 475 mL 475mL
11976079 ISA Solution Bromide Electrode, Cadmium Electrode, Chloride Electrode, Cupric Electrode, Iodide Electrode, Sufide Electrode, Thiocyanate Electrode 475 mL 475mL
11757647 TISAB II Fluoride Electrode 1 Gal. 3.8 L
11966089 TISAB III Fluoride Electrode 475 mL 475mL
10665261 TISAB II Fluoride Electrode 4 x 1 Gal. Pack of 4
11777777 Sulfide Anti-Oxidant Buffer Sulfide Electrode 4 x 475 mL 475mL
11747757 CISA Chloride Electrode 2 x 1 L Pack of 2
10598741 ISA Solution Carbon Dioxide Electrode 475 mL 475mL
11777677 Alkaline Reagent Cyanide Electrode 475 mL 475mL
12993247 pH Adjusting Low Level ISA Ammonia Electrode 475 mL 475mL
12881521 pH Adjusting ISA Ammonia Electrode 475 mL 475mL
12903257 Storage Solution Ammonia Electrode 475 mL 475mL
11767687 Acid Buffer Nitrogen Oxide Electrode 475 mL 475mL
11767807 Iodide Reagent Chlorine Electrode 5 x 50 mL Pack of 5
11737707 Acid Reagent Chlorine Electrode 475 mL 475mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11777527 Fournisseur Thermo Scientific™ Code fournisseur 930711
  • ONLINE EXCLUSIVE PRICE
    111.56 €
    105.00 € / 475mL
    Save 6.56 €
    5% Off
Afficher toutes les promotions
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents.

Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents. Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.

Warranty: Out of box

Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements. The ammonia pH-adjusting ISA (951211) and low-level ammonia pH-adjusting ISA (951210) both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required. The ammonia pH-adjusting ISA (951211) prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results. The low-level ammonia pH-adjusting ISA (951210) is ideal for ammonia samples with a concentration of 10ppm and below that do not contain metal interferences.

All ionic strength adjusters and special reagents are packaged as one 1 pint (475mL) bottle, except:

  • Cat. No. 941709 is packaged as two 1 quart (2 x 1L) bottles
  • Cat. No. 941609 is packaged as four 1 pint (4 x 475mL) bottles
  • Cat. No. 940909 is packaged as one 1 gallon (3.7L) bottle
  • Cat. No. 940999 is packaged as four 1 gallon (4 x 3.7L) bottles
TRUSTED_SUSTAINABILITY

Spécification

Description ISA for Fluoroborate, Nitrate, Perchlorate Electrodes
For Use With (Equipment) Fluoroborate Electrode, Nitrate Electrode, Perchlorate Electrode
Type ISA Solution
For Use With (Application) Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes
Packaging Bottle
Quantity 475 mL
compliance-icons
Identifiant produit
  • Nitrate Ionic Strength Adjustor
Catégorie de danger
  • LONG-TERM AQUATIC HAZARD Chronic 2
Mention de danger
  • H411-Toxic to aquatic life with long lasting effects.
Conseils de prudence
  • P202-Do not handle until all safety precautions have been read and understood.
  • P273-Avoid release to the environment.
Informations supplémentaires
  • MIXTURE LIST-Contain: Ammonium Sulfate

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.