missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents

Product Code. 12903257
Change view
Click to view available options
Type:
Acid Buffer
Acid Reagent
Alkaline Reagent
CISA
ISA Solution
Iodide Reagent
Ionic Strength Adjustor Buffer
Nitrate Interference Suppressor
Storage Solution
Sulfide Anti-Oxidant Buffer
TISAB II
TISAB III
pH Adjusting ISA
pH Adjusting Low Level ISA
For Use With (Equipment):
Ammonia Electrode
Bromide Electrode, Cadmium Electrode, Chloride Electrode, Cupric Electrode, Iodide Electrode, Sufide Electrode, Thiocyanate Electrode
Calcium Electrode
Carbon Dioxide Electrode
Chloride Electrode
Chlorine Electrode
Cyanide Electrode
Fluoride Electrode
Fluoroborate Electrode, Nitrate Electrode, Perchlorate Electrode
Nitrate Electrode
Nitrogen Oxide Electrode
Potassium Electrode
Sodium Electrode
Sodium ISE
Sulfide Electrode
Quantity:
475 mL
5 x 50 mL
1 Gal.
4 x 475 mL
2 x 1 L
4 x 1 Gal.
Unit Size:
3.8 L
475mL
Each
Pack of 2
Pack of 4
Pack of 5
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Quantity Unit Size
12903257 Storage Solution Ammonia Electrode 475 mL 475mL
11722787 Storage Solution Sodium Electrode 475 mL Each
12049967 Ionic Strength Adjustor Buffer Sodium ISE 475 mL 475mL
11767527 Nitrate Interference Suppressor Nitrate Electrode 475 mL 475mL
11777527 ISA Solution Fluoroborate Electrode, Nitrate Electrode, Perchlorate Electrode 475 mL 475mL
11717537 ISA Solution Potassium Electrode 475 mL 475mL
11747537 ISA Solution Calcium Electrode 475 mL 475mL
11976079 ISA Solution Bromide Electrode, Cadmium Electrode, Chloride Electrode, Cupric Electrode, Iodide Electrode, Sufide Electrode, Thiocyanate Electrode 475 mL 475mL
11757647 TISAB II Fluoride Electrode 1 Gal. 3.8 L
11966089 TISAB III Fluoride Electrode 475 mL 475mL
10665261 TISAB II Fluoride Electrode 4 x 1 Gal. Pack of 4
11777777 Sulfide Anti-Oxidant Buffer Sulfide Electrode 4 x 475 mL 475mL
11747757 CISA Chloride Electrode 2 x 1 L Pack of 2
10598741 ISA Solution Carbon Dioxide Electrode 475 mL 475mL
11777677 Alkaline Reagent Cyanide Electrode 475 mL 475mL
12993247 pH Adjusting Low Level ISA Ammonia Electrode 475 mL 475mL
12881521 pH Adjusting ISA Ammonia Electrode 475 mL 475mL
11767687 Acid Buffer Nitrogen Oxide Electrode 475 mL 475mL
11767807 Iodide Reagent Chlorine Electrode 5 x 50 mL Pack of 5
11737707 Acid Reagent Chlorine Electrode 475 mL 475mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 12903257 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' 951213
  • ONLINE EXCLUSIVE PRICE
    79.09 €
    66.15 € / 475mL
    Save 12.94 €
    16% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents.

Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents. Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.

Warranty: Out of box

Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements. The ammonia pH-adjusting ISA (951211) and low-level ammonia pH-adjusting ISA (951210) both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required. The ammonia pH-adjusting ISA (951211) prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results. The low-level ammonia pH-adjusting ISA (951210) is ideal for ammonia samples with a concentration of 10ppm and below that do not contain metal interferences.

All ionic strength adjusters and special reagents are packaged as one 1 pint (475mL) bottle, except:

  • Cat. No. 941709 is packaged as two 1 quart (2 x 1L) bottles
  • Cat. No. 941609 is packaged as four 1 pint (4 x 475mL) bottles
  • Cat. No. 940909 is packaged as one 1 gallon (3.7L) bottle
  • Cat. No. 940999 is packaged as four 1 gallon (4 x 3.7L) bottles
TRUSTED_SUSTAINABILITY

Specifications

Description Storage Solution for Ammonia Electrode
For Use With (Equipment) Ammonia Electrode
Type Storage Solution
For Use With (Application) Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes
Packaging Bottle
Quantity 475 mL

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.