missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion Lab Star PH111 Bench pH Meters

Product Code. 17731849
Change view
Click to view available options
Type:
pH/mV Meter Bio Kit
pH/mV Meter Difficult Sample Kit
pH/mV Meter Low Main. Bio Kit
pH/mV Meter Standard Kit
pH/mV Meter with Basic Electrode
pH/mV Meter with Stand
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
17731849 pH/mV Meter with Stand Each
17741849 pH/mV Meter Standard Kit Each
17751849 pH/mV Meter Bio Kit Each
17761849 pH/mV Meter Low Main. Bio Kit Each
17771849 pH/mV Meter Difficult Sample Kit Each
17781849 pH/mV Meter with Basic Electrode Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17731849 Supplier Thermo Scientific™ Supplier No. LSTAR1110
  • ONLINE EXCLUSIVE PRICE
    687.37 €
    596.11 € / Pack of 1
    Save 91.26 €
    13% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Orion Lab Star PH111 benchtop pH meters measure pH or ORP with mV and temperature, feature a large backlit display for easy viewability, and offer small footprint to save bench space. The Orion Lab Star pH meter is ideal for simplified, budget-friendly lab testing and data recording.

Discover the award-winning Orion Lab Star PH111 pH/mV bench meter. Measure pH or ORP simultaneously with mV and temperature. Featuring a large, backlit display for easy viewability and small footprint to save bench space, this pH meter is ideal for simplified, budget-friendly lab testing. Use the onscreen stabilizing and ready indicators to help identify when pH meter and ORP meter readings are complete. View active pH, ORP and temperature calibration logs with date and time stamp, record key measurements using the 500-point data log with date and time stamp, and output all logs to a printer or computer.

Orion Lab Star PH111 pH Meter Features

  • Budget-friendly pH meter for simplified pH and ORP testing
  • Display shows pH, mV and temperature simultaneously for accurate data recording
  • Measure -2.000 to 18.000 pH, -2000.0 to +2000.0 mV and -5.0 to 105.0 °C or 23.0 to 221.0 °F to ensure comprehensive measurement capabilities
  • Perform one to five point pH calibrations, one point ORP calibrations and one point temperature adjustment calibrations for proficient SOP compliance
  • Large 5 inch backlit display makes viewing and recording measurements easy
  • Small meter footprint saves valuable bench space in your lab
  • Onscreen stabilizing and ready indicators help identify when readings are complete
  • pH electrode condition icon helps ensure reliable electrode performance
  • Date and time data stamped data and calibration logs support GLP compliance
  • View active pH, ORP and temperature calibration logs with date and time stamp to ensure accurate, reproducible measurements
  • Use 500-point data log with date and time stamp to record key measurements
  • Output data and calibration logs to a printer or computer for long-term data storage

Meter and Meter Kits

  • LSTAR1110 is the pH meter and stand option, which allows you to purchase your pH electrode separately
  • LSTAR1115 is the standard pH meter kit option, for common pH samples such as drinking water, tap water, buffers (non-TRIS) and general purpose student use
  • LSTAR116 is the easy-to-clean bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included refillable pH electrode is easy-to-clean and restore
  • LSTAR117 is the low-maintenance bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included gel-filled pH electrode is low-maintenance and easy to operate
  • LSTAR1118 is the difficult sample pH meter kit option, for pH samples that are thick, viscous or semi-solid including foods, beverages, wastewaters, emulsions, soils, slurries, suspended solids or sludges
TRUSTED_SUSTAINABILITY

Specifications

Description Orion Lab Star PH111 bench pH/mV meter with stand
Includes Meter with electrode stand, PC cable, universal power adapter
Accuracy (Temperature) ±0.3°C/±0.5°F
Accuracy (mV) ±0.2 or ±0.05%, whichever is greater
Accuracy (pH) ±0.005, ±1 LSD
Data Management Details Transfer single or all data points to printer or computer
Outputs Computer or printer (STARA-106)
For Use With (Application) Meter and Stand Option, Select and Purchase The Electrode Separately
Measurement Modes pH with mV and Temperature, ORP with mV and Temperature
Calibration pH, ORP and temperature
Range (pH) -2 to 18
Resolution (pH) 0.001, 0.01, 0.1
Calibration Points (pH) 1 to 5 points
Range (mV) ±2000.0
Range (ORP) ±2000.0 mV with ±250 mV offset
Resolution (mV) 0.1 mV
Calibration Points 1 (ORP)
Temperature Range (Metric) -5.0°C to +105.0°C
Temperature Range (English) 23.0°F to 221.0°F
Resolution (Temperature) 0.1°C, 0.1°F
Temperature Selection Manual or automatic with ATC temperature probe
Temperature Probe Calibration 1 point temperature offset calibration
Display Type LCD, 5 in. Backlit
Backlight Option Yes
Memory Non-volatile
Data Points 500 with Date and Time Stamp
Log Function Type Automatic with Auto-Read and Timed Read Modes, Manual with Continuous Read Mode
Alarm Output Calibration reminder alarm
Inputs BNC for pH or ORP, 8-pin MiniDIN for ATC
Certifications/Compliance CE, TUV 3-1, FCC Class A
IP Rating IP54
AC Adapter Universal 100-240 V 50/60 Hz power supply with twist-and-lock meter connection and interchangeable wall plugs for US/Japan, Euro, UK/Singapore, Australia/New Zealand and China outlets
Dimensions (L x W x H) 198.3 x 155.2 x 61.7 mm, 7.81 x 6.11 x 2.43 in.
Weight (Metric) 700 grams
Weight (English) 1.54 lb.
Warranty 3 year meter warranty
Type pH/mV Meter with Stand
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.