Learn More
Thermo Scientific™ Orion™ Ionplus™ Sure-Flow™ Lead Combination Ion Selective Electrode
Product Code. 10587401
Change view
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
Screw Cap
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
10587401 BNC (Waterproof) Each
10191251 BNC Each
10479102 Screw Cap Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10587401 Supplier Thermo Scientific™ Supplier No. 9682BNWP
  • ONLINE EXCLUSIVE PRICE
    1145.70 €
    909.00 € / Pack of 1
    Save 236.70 €
    20% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Measure lead ions in aqueous solutions quickly, simply, accurately and economically.
Measure lead ions in aqueous solutions quickly, simply, accurately and economically with Thermo Scientific™ Orion™ Lead Electrodes.
Features
Free-flowing Themo Scientific™ Sure-Flow™ junction assures stable, drift-free potentials. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap The lead electrode makes a sensitive endpoint detector for the titration of sulfate ions with lead perchlorate.
Solutions required:
Standard
948206 – 0.1 M lead standard, 475 mL bottle
948207 – 0.1 M sodium sulfate standard for sulfateanalysis with the lead ISE, 475 mL bottle
Electrode Fill Solution
900062 – Optimum Results B electrode filling solution, 5 x 60 mL bottles
Items included
900062 – Optimum Results B electrode filling solution, 60 mL 1 bottle
948201 – Polishing strip. Qty 2.
Warranty period
12 months from date of purchase
TRUSTED_SUSTAINABILITY
Specifications
Connector Types BNC (Waterproof)
Description With waterproof BNC connector
Drift drift-free potentials
Junction Sure-Flow Junction
For Use With (Equipment) ISE Meter
Type Lead Ion Selective Electrode, BNC waterproof connector