missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions

Product Code. 11727777
Change view
Click to view available options
Filling Solution Type:
A, B, C, D
Bacteria Removal (B)
No Cal Stain Lifter
Oil and Grease Removal (D)
Protein Removal (A)
Rinse
Quantity:
10 pk.
25 pk.
4 x 30 mL
4 x 60 mL
60 mL
60 mL (B, D), 30 mL (A, C)
Unit Size:
60mL
Pack of 10
Pack of 25
Pack of 4
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filling Solution Type Quantity Unit Size
11727777 Protein Removal (A) 4 x 30 mL Pack of 4
11722737 No Cal Stain Lifter 60 mL 60mL
11772787 A, B, C, D 60 mL (B, D), 30 mL (A, C) Pack of 4
15304338 Protein Removal (A) 4 x 30 mL Pack of 4
11737777 Bacteria Removal (B) 4 x 60 mL Pack of 4
11757777 Oil and Grease Removal (D) 4 x 60 mL Pack of 4
11772168 Rinse 10 pk. Pack of 10
11702188 Rinse 25 pk. Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 11727777 Supplier Thermo Scientific™ Supplier No. 900021WA
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Restore proper electrode performance and prolong electrode life using these pH electrode cleaning solutions.

Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions in ready to use bottles.

Cleaning Solutions:

  • Solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation
  • Cleaning solution A is for removing protein contaminants, cleaning solution B is for removing bacterial contaminants, cleaning solution C is for general cleaning and cleaning solution D is for removing oil and grease contaminants
  • Bottles include a small beaker to hold the solution and a plastic pipet to remove the old electrode filling solution

Includes (PN 900020):

  • One bottle each of cleaning solutions A, B, C and D
  • One beaker
  • One pipet

Note:

The following part numbers have been changed due to a recent system upgrade:

Old Part Number: 900021

New Part Number: 900021-WA

Old Part Number: 900022

New Part Number: 900022-WA

Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.

Warranty: Out of box

TRUSTED_SUSTAINABILITY

Specifications

Description 4 x 1 oz. (4 x 30mL) Bottles of Cleaning Solution A for Protein Removal, Beaker and Pipet
For Use With (Equipment) pH Electrode
Type Cleaning Solution
Filling Solution Type Protein Removal (A)
For Use With (Application) Solutions restore proper electrode performance and prolong the useful life of the electrode
Packaging Bottle
Quantity 4 x 30 mL
compliance-icons
Product Identifier
  • pH Cleaner A
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH208-Contains . May produce an allergic reaction.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P321-Specific treatment (see supplemental first aid instructions on this label).
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P202-Do not handle until all safety precautions have been read and understood.
Supplemental information
  • MIXTURE LIST-Contain: Pepsin, Hydrochloricacid

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.