Learn More
Thermo Scientific™ Orion™ Potassium Ion Selective Electrodes
Product Code. 10282871
Change view
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
None
For Use With (Equipment):
Combination Electrode
Half-Cell Electrode
ISE Meter
Type:
Combination Electrode
Half Cell Electrode
Replacement Module
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types For Use With (Equipment) Type unitSize
10282871 None Half-Cell Electrode Replacement Module Each
10410643 BNC ISE Meter Half Cell Electrode Each
10390832 None Combination Electrode Replacement Module Each
10757352 BNC (Waterproof) ISE Meter Combination Electrode Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10282871 Supplier Thermo Scientific™ Supplier No. 931901
  • ONLINE EXCLUSIVE PRICE
    278.00 €
    227.70 € / Pack of 1
    Save 50.30 €
    18% Off
View all promotions
In Stock
Add to basket
Add to basket
Measure potassium ions in aqueous solutions simply, accurately and economically.
Measure potassium ions in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Potassium Electrodes.
  • Calibration Standard Catalog number: 0.1M KCl / 921906
  • Required ISA Catalog number: 931911
TRUSTED_SUSTAINABILITY
Specifications
Connector Types None
Description Replacement Module for Potassium Half-Cell Electrode 9319BN
Drift drift-free potentials
Junction Sure-Flow Junction
For Use With (Equipment) Half-Cell Electrode
Type Replacement Module
Parameter Potassium