missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion Star™ A326 pH/Dissolved Oxygen Portable Multiparameter Meter
BIOPHARMA

Product Code. 11691479
Change view
Click to view available options
Product Type:
Portable Meter
Portable Meter Kit
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11691479 Portable Meter Each
11601489 Portable Meter Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11691479 Supplier Thermo Scientific™ Supplier No. STARA3260
  • ONLINE EXCLUSIVE PRICE
    1400.00 €
    1330.00 € / Pack of 1
    Save 70.00 €
    5% Off
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accurately measure pH, dissolved oxygen and temperature with our pH/DO/RDO portable meter for high performance in challenging field tests.

Get high accuracy and premium performance everywhere you need it with the Thermo Scientific™ Orion Star™ A326 pH/Dissolved Oxygen Portable Multiparameter Meter. This meter is designed for a wide range of pH, mV, ORP, dissolved oxygen and temperature testing and field applications. Evaluate samples in the most demanding locations using this waterproof meter with IP67-rated housing. Use the two meter channels to measure pH and dissolved oxygen simultaneously or view each channel separately. Log up to 5000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer.

Take your measurements at the source with our premium line of portable meters. The Thermo Scientific Orion Star A326 pH/RDO/DO Portable Meter offers an information-rich screen and user-friendly operation. This rugged, waterproof portable meter is ready to tackle the elements with you. Orion Star A326 pH/RDO/DO Portable Meters are designed to be easy-to-use, dependable field meters that offer advanced features and functions including a 5000 point date log with sample ID and user ID options, calibration and high/low limit alarms and password-protected measurement methods.

  • Combined convenience with the benefits of a pH meter and dissolved oxygen meter in one easy-to-use system
  • Measure pH, mV, relative mV or ORP with temperature on channel one and measure dissolved oxygen in percent saturation or mg/L with temperature using RDO optical DO probes or polarographic DO probes on channel two
  • Easily view individual measurements on a full screen display and quickly toggle to a split screen display to view multiple measurements simultaneously
  • Perform up to a 5 point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values and calibration editing option that allows individual points to be fixed without a full recalibration
  • Calibrate dissolved oxygen using water-saturated air, air-saturated water, custom value (Winkler titration) and zero point options
  • Automatically compensate dissolved oxygen readings using ATC temperature, manual salinity input and built-in barometer for barometric pressure readings
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Capture your critical readings with measurement stability indicator and selectable read modes — Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
  • Data log collects and date/time stamps up to 5,000 measurement sets with optional sample ID and user ID
  • Calibration log stores 10 most recent calibrations
  • Non-volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Flexible power options with AA batteries or optional universal power adapter
  • IP67-rated housing is waterproof for rugged field operation
  • Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
  • Meters offer CE, TUV 3-in-1 and FCC Class A certifications
  • 3-year replacement warranty
TRUSTED_SUSTAINABILITY

Specifications

AC Adapter Universal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003)
Outputs USB, RS232
Accuracy (Barometer) ±6mmHg
Accuracy (mV) ±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH) ±0.002
Accuracy (Polarographic DO) ±0.2mg/L or ±2% of reading, whichever is greater
Accuracy (RDO) ±0.1mg/L up to 8mg/L, ±0.2mg/L from 8 to 20mg/L, ±10% of reading up to 50mg/L
Alarm Output High/low limit alarm, calibration due alarm
Backlight Option Yes (Selectable)
Barometric Pressure Correction 400 to 850mmHg, automatic using built-in barometer or manual entry
Battery Life 800 hr.
Battery Type 4 AA (included)
Calibration pH with calibration editing option, relative mV (RmV), ORP, dissolved oxygen and temperature
Calibration Modes Water-saturated air, air-saturated water, manual (Winkler titration), zero point
Calibration Points 1 point relative mV (RmV) mode, 1 point ORP mode
Calibration Points (pH) 1 to 5
Certifications/Compliance CE, TUV 3-1, FCC Class A
Data Management Details Transfer single, range or all data points to printer or computer
Data Points 5000 with Date and Time Stamp
Description Orion Star A326 pH/RDO/DO portable meter
Channels 2
Display Type LCD Graphic, Backlit
IP Rating IP67
Measurement Modes pH, mV, Relative mV, ORP with Temperature, Dissolved Oxygen (% Saturation), Dissolved Oxygen (mg/L with Temperature)
Percent Saturation Range (Polarographic DO) 0.0 to 600.0 % saturation
Range (mV) ±2000.0 mV
Range (pH) -2 to 20
Resolution (pH) 0.1/0.01/0.001
Temperature Range (Metric) -5.0°C to +105.0°C
Includes Star A326 pH/RDO/DO meter, 4 AA batteries, literature CD, printed quick start guide, computer interface cable and meter test certificate
Inputs BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature)
Isopotential Point 7.000pH
Keypad Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
Languages English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
Log Function Type Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m
Memory Non-volatile memory preserves data log, calibration log and meter settings
Methods 10 per channel with password protection
Model Orion Star A326 pH/mV/RmV/ORP/RDO Optical/Dissolved Oxygen/Temperature Portable Meter
Percent Saturation Relative Accuracy (RDO) ±2 % saturation ≤ 200 % saturation, ±10 % saturation > 200 % saturation
Percent Saturation Resolution (Polarographic DO) 0.1, 1 % saturation
Percent Saturation Range (RDO) 0.0 to 500.0 % saturation
Percent Saturation Relative Accuracy (Polarographic DO) ±2 % saturation or ±2% of reading whichever is greater
Percent Saturation Resolution (RDO) 0.1, 1 % saturation
Power Supply 4 AA batteries (included) or universal AC adapter (sold separately)
Pressure Range 400 to 850mmHg
Pressure Resolution 1mmHg
Product Type Portable Meter
Range (ORP) ±2000.0 mV
Range (Polarographic DO) 0.00 to 90.00mg/L
Range (RDO) 0.00 to 50.00mg/L
Range (Relative mV) ±2000.0 mV
Relative Humidity Range 5 to 85 %, non-condensing
Resolution (mV) 0.1mV
Resolution (Polarographic DO) 0.01, 0.1mg/L
Resolution (RDO) 0.01, 0.1mg/L
Salinity Factor 0.0 to 45.0 ppt, automatic correction using manual entry of sample salinity
Stability Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
Temperature Accuracy ±0.1°C
Temperature (English) Operating 41°F to 113°F (ambient)
Temperature (Metric) Operating 5°C to 45°C (ambient)
Temperature Probe Calibration 1 point temperature offset calibration
Temperature Range (English) 22.0°F to 221.0° F
Temperature Resolution 0.1°C, 0.1°F
Temperature Selection Manual or automatic with ATC temperature probe
Warranty 3 Year Meter
Dimensions (L x W x H) 24cm x 10cm x 6.5cm (9.5 in. x 3.9 in. x 2.6 in.)
Weight (English) 1.2 lb.
Weight (Metric) 0.5 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.