missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Versa Star pH Measurement Module

Artikelnummer. 11621519
Ansicht ändern
Click to view available options
pH Range:
-2.000 to 20.000
Packungsgröße:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. pH Range Packungsgröße
11621519 -2.000 to 20.000 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11621519 Lieferant Thermo Scientific™ Lieferanten-Nr. VSTARPH
  • ONLINE EXCLUSIVE PRICE
    597.98 €
    476.10 € / Pack of 1
    Save 121.88 €
    20% Off
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Add pH, mV, ORP and temperature measurement capabilities to your Versa Star meter using the Thermo Scientific™ Orion™ Versa Star pH Measurement Module

Add pH, mV, ORP and temperature measurement capabilities to your Thermo Scientific™ Orion™ Versa Star meter using the Thermo Scientific™ Orion™ Versa Star pH Measurement Module. Each Orion™ Versa Star meter has four channels that can be customized with interchangeable measurement modules. The Versa Star pH module offers up to a six point pH calibration with easy to follow onscreen instructions, automatic buffer recognition of NIST, DIN or user-defined buffer groups and calibration editing option to fix individual points without a full recalibration.

Select from five Thermo Scientific Orion Versa Star measurement modules to customize the measurement capabilities of your Orion Versa Star meter. Measurement modules are interchangeable and can be added or removed from each Versa Star meter's four channels at any time.

Orion Versa Star pH Measurement Module:

  • Measure pH, mV, relative mV or ORP with temperature
  • Perform up to a six point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Customize measurements to show calibration history and pH electrode condition or simplify to show an extra large pH reading
  • Module offers a 3-year replacement warranty

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

TRUSTED_SUSTAINABILITY

Spezifikation

Accuracy (pH) ±0.002 pH
Accuracy (mV) ±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (Temperature) ±0.1°C
Alarm Output Set point alarm, high/low limit alarm, calibration due alarm
Calibration pH with calibration editing option, relative mV (RmV), ORP and temperature
Calibration Points 1 to 6
Description Orion Versa Star pH measurement module
Type pH Measurement Module
Dimensions (L x W x H) 12 x 3 x 9.5 cm (4.7 x 1.2 x 3.8 in.)
Inputs BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
Includes Versa Star pH measurement module for use with Versa Star Pro meters to measure pH, mV, relative mV, ORP and temperature using ATC probe or manual entry
Isopotential Point 7.000pH (adjustable)
IP Rating IP-54
Measurement Modes pH, mV, relative mV (RmV) or ORP with temperature
Methods 10 per channel with password protection
Model Orion Versa Star pH/mV/RmV/ORP/Temperature Measurement Module
pH Range -2.000 to 20.000
Probe Type pH electrode, ORP electrode, reference half-cell electrode, ATC temperature probe (purchase separately)
Range (mV) ±2000.0 mV
Range (ORP) ±2000.0mV
Relative Humidity Range 5 to 85%, non-condensing
Resolution (mV) 0.1mV
Resolution (pH) 0.1, 0.01, 0.001
Resolution (Temperature) 0.1°F/°C
Stability Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
Temperature (English) Range 22.0°F to 221.0°F
Temperature (English) Operating 41°F to 113°F
Temperature (Metric) Range -5°C to 105deg;C
Temperature Probe Calibration 1 point temperature offset calibration
Temperature (Metric) Operating 5°C to 45°C
Temperature Selection Manual or automatic with ATC temperature probe
Weight (English) 0.5 lb.
Warranty 3 year module warranty
Weight (Metric) 0.2 kg
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.