missing translation for 'onlineSavingsMsg'
Learn More

Ornidazole, 99%

Product Code. 15435369
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435369 25 g 25g
15425369 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435369 Supplier Thermo Scientific Alfa Aesar Supplier No. J63100.14
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Interferes with electron transport in metabolic pathways | CAS: 16773-42-5 | C7H10ClN3O3 | 219.625 g/mol

Ornidazole is a drug that cures some protozoan infections. It has been investigated for use in Crohn's disease after bowel resection. It is indicated in treatment of mixed amoebiasis, mixed amoebic dysentery, mixed giardiasis, trichomoniasis, bacterial vaginosis, Sexually transmitted diseases, infections of gynaecology, lower respiratory tract infections, ENT, surgical and dental infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethanol at 50mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16773-42-5
Molecular Formula C7H10ClN3O3
Molecular Weight (g/mol) 219.625
MDL Number MFCD00057960
InChI Key IPWKIXLWTCNBKN-UHFFFAOYSA-N
Synonym ornidazole, madelen, tiberal, ornidazolum, 1-3-chloro-2-hydroxypropyl-2-methyl-5-nitroimidazole, ornidazol, 1-chloro-3-2-methyl-5-nitro-1h-imidazol-1-yl propan-2-ol, 1-chloro-3-2-methyl-5-nitro-1h-imidazol-1-yl-2-propanol, ornidal, 1-2-hydroxy-3-chloropropyl-2-methyl-5-nitroimidazole
PubChem CID 28061
ChEBI CHEBI:75176
IUPAC Name 1-chloro-3-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol
SMILES CC1=NC=C(N1CC(CCl)O)[N+](=O)[O-]

Specifications

Melting Point 84°C to 86°C
Color White to Yellow
Quantity 25 g
Merck Index 14,6872
Solubility Information Soluble in ethanol at 50mg/ml
Formula Weight 219.63
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Ornidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.