missing translation for 'onlineSavingsMsg'
Learn More

Orthophosphoric Acid, 85+%, Certified AR for Analysis, Fisher Chemical™

Product Code. 10559000 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic Coated Glass bottle
Plastic container
Unit Size:
1 L
2.5 L
25 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10559000 2.5 L HDPE plastic bottle 2.5 L
10244010 500 mL HDPE plastic bottle 500mL
10438990 1 L HDPE plastic bottle 1 L
11478443 2.5 L Plastic Coated Glass bottle 2.5 L
12646747 25 L Plastic container 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10559000 Supplier Fisher Chemical Supplier No. O/0500/PB17
  • ONLINE EXCLUSIVE PRICE
    123.17 €
    105.00 € / 2.5 L
    Save 18.17 €
    14% Off
View all promotions
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

H3O4P, CAS Number-7664-38-2, acide phosphorique, acido fosforico, acidum phosphoricum, evits, o-phosphoric acid, orthophosphoric acid, phosphorsaeure, sonac, wc-reiniger, white, 2.5L, 158 deg.C, CHEBI:26078, Undesignated, NBIIXXVUZAFLBC-UHFFFAOYSA-N, 21 deg.C, 11340, 97.994, HDPE plastic bottle | CAS: 7664-38-2 | H3O4P | 97.994 g/mol

H3O4P, CAS Number-7664-38-2, acide phosphorique, acido fosforico, acidum phosphoricum, evits, o-phosphoric acid, orthophosphoric acid, phosphorsaeure, sonac, wc-reiniger, white, 2.5L, 158 deg.C, CHEBI:26078, Undesignated, NBIIXXVUZAFLBC-UHFFFAOYSA-N, 21 deg.C, 11340, 97.994, HDPE plastic bottle

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number 11340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Boiling Point 158°C
Melting Point 21°C
pH 2
Physical Form Liquid
Quantity 2.5 L
Packaging HDPE plastic bottle
Vapor Pressure 2 hPa at 20°C
Density Vapor Density: 3.4
Chemical Name or Material Orthophosphoric Acid
compliance-icons
Product Identifier
  • Phosphoric acid, 85+% solution in water
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Orthophosphoric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.