missing translation for 'onlineSavingsMsg'
Learn More

Oseltamivir phosphate, 98%, Thermo Scientific Chemicals

Product Code. 15698715 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15698715 1 g 1g
15608725 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15698715 Supplier Thermo Scientific Acros Supplier No. 461170010
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 204255-11-8
Molecular Formula C16H28N2O4·H3PO4
Molecular Weight (g/mol) 410.4
InChI Key PGZUMBJQJWIWGJ-ONAKXNSWSA-N
Synonym oseltamivir phosphate, tamiflu, oseltamir phosphate, unii-4a3o49ngez, 4a3o49ngez, ethyl 3r,4r,5s-4-acetamido-5-amino-3-pentan-3-yloxy cyclohex-1-ene-1-carboxylate phosphate, ethyl 3r,4r,5s-4-acetamido-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1, osteltamivir phosphate, 3r-3alpha,4beta,5alpha-ethyl 4-acetylamino-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1
PubChem CID 78000
ChEBI CHEBI:7799
IUPAC Name ethyl (3R,4R,5S)-4-acetamido-5-amino-3-pentan-3-yloxycyclohexene-1-carboxylate;phosphoric acid
SMILES CCC(CC)OC1C=C(CC(C1NC(=O)C)N)C(=O)OCC.OP(=O)(O)O

Specifications

Color White to Off-White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 410.4
Percent Purity 98%
Chemical Name or Material Oseltamivir phosphate
compliance-icons
Product Identifier
  • Oseltamivir phosphate, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Oseltamivir phosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.