missing translation for 'onlineSavingsMsg'
Learn More

Osmium(III) chloride hydrate

Product Code. 10450351 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10450351 500 mg Glass bottle 500mg
10721555 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10450351 Supplier Thermo Scientific Acros Supplier No. 318705000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14996-60-2 | Cl3H2OOs | 314.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14996-60-2
Molecular Formula Cl3H2OOs
Molecular Weight (g/mol) 314.60
MDL Number MFCD00149815
InChI Key KHHOLOMEVBPRQY-UHFFFAOYSA-K
Synonym osmium iii chloride hydrate, osmium chloride oscl3 , hydrate 8ci,9ci, trichloroosmium hydrate, cl3os.h2o, trichloroosmium-water 1/1
PubChem CID 16211485
SMILES O.Cl[Os](Cl)Cl

Specifications

Melting Point >500.0°C
Color Black to Red
Physical Form Crystals or Powder
Quantity 500 mg
Linear Formula OsCl3·xH2O
Solubility Information Solubility in water: soluble
IUPAC Name trichloroosmium;hydrate
Formula Weight 296.56
Packaging Glass bottle
Chemical Name or Material Osmium(III) chloride hydrate
compliance-icons compliance-icons
Product Identifier
  • Osmium(III) chloride hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Osmium(III) chloridehydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.