missing translation for 'onlineSavingsMsg'
Learn More

Osmium(III) chloride trihydrate, Premion™, 99.99% (metals basis), Os 52-56%

Product Code. 11307738
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11307738 5 g 5g
11317738 0.25 g 0.25g
11327738 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11307738 Supplier Thermo Scientific Alfa Aesar Supplier No. 033480.06
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 135296-80-9 | Cl3H6O3Os | 350.625 g/mol

Osmium(III) chloride trihydrate is used as a reagent for preparation of dichlorodihydridoosmium complexes. It is also useful as starting materials and an efficient alkane oxidation catalyst in the presence of pyridine and hydrogen oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135296-80-9
Molecular Formula Cl3H6O3Os
Molecular Weight (g/mol) 350.625
MDL Number MFCD00011148
InChI Key UACQLNPCDXDCID-UHFFFAOYSA-K
Synonym osmium iii chloride trihydrate, osmium trichloride trihydrate, cl3os.3h2o, acmc-20ak04, trichloroosmium-water 1/3, trichloroosmium trihydrate, osmium iii chloride trihydrate, premion
PubChem CID 57348105
IUPAC Name trichloroosmium;trihydrate
SMILES O.O.O.Cl[Os](Cl)Cl

Specifications

Melting Point (decomposition)
Color Black-Red
Physical Form Crystalline
Quantity 5 g
Linear Formula OsCl3·3H2O
UN Number UN3260
Solubility Information Soluble in water.
Formula Weight 350.60 (296.56 Anhydrous)
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Os 52 to 56%
Assay (metals basis)
Chemical Name or Material Osmium(III) chloride trihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.