missing translation for 'onlineSavingsMsg'
Learn More

Osmium Tetroxide, ACS Reagent, ≥98%, Honeywell Fluka™

Product Code. p-7120640 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
15625480 250 mg Glass Bottle 250mg
15615480 1 g Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 15625480

Brand: Honeywell-Fluka 419494250MG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O

Specifications

Boiling Point 130°C
Melting Point 39.5°C to 41°C
Physical Form Solid
Quantity 250 mg
Linear Formula OsO4
UN Number UN2471
Solubility Information Slightly soluble in water
Formula Weight 254.23g/mol
Percent Purity ≥98%
Odor Unpleasant
Packaging Glass Bottle
Vapor Pressure 7mmHg (20°C)
Density Vapor Density: 8.8 (vs air)
Chemical Name or Material Osmium tetroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.