missing translation for 'onlineSavingsMsg'
Learn More

Osmium(VIII) oxide, 2% aq. soln.

Product Code. 15442237
Change view
Click to view available options
Quantity:
2 mL
10 mL
Unit Size:
10mL
2mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15442237 2 mL 2mL
15452237 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15442237 Supplier Thermo Scientific Alfa Aesar Supplier No. 045384.9W
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20816-12-0 | O4Os | 254.23 g/mol

Osmium(VIII) oxide is widely used as an oxidant in organic synthesis to oxidize alkenes to vicinal diols as well as in the sharpless oxyamination to give vicinal amino-alcohols. It is involved in the oxidative cleavage of alkenes (Lemieux-Johnson oxidation) in association with sodium periodate. It finds application in transmission electron microscopy (TEM) as a staining agent. It is involved in the promotion of oxidative cleavage of graphene layers in carbon nanotubes and as an intermediate in osmium ore refining. As a lipid stain, it is useful in scanning electron microscopy (SEM) as an alternative to sputter coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O

Specifications

Color Colorless to Yellow
Physical Form Liquid
Quantity 2 mL
UN Number UN3287
Merck Index 14,6893
Solubility Information Soluble in alcohol,ether,chloroform,benzene,ammonium hydroxide,phosphorus oxychloride and carbon tetrachloride
Formula Weight 254.1
Concentration or Composition (by Analyte or Components) 2% aq. soln.
Odor Chlorine-like
Sensitivity Air Sensitive
Chemical Name or Material Osmium (VIII) oxide
Show More Show Less
compliance-icons
Product Identifier
  • Osmium(VIII) oxide, 2% aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Osmium(VIII) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.