missing translation for 'onlineSavingsMsg'
Learn More

Osthole, MedChemExpress

Product Code. 16451288
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451288 5 g 5g
16449895 250 mg 250mg
16442489 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16451288 Supplier Medchem Express Supplier No. HYN0054
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Osthole (Osthol) is a natural antihistamine alternative. Osthole may be a potential inhibitor of histamine H1 receptor activity. Osthole also suppresses the secretion of HBV in cells.

Osthole (Osthol) is a natural antihistamine alternative. Osthole may be a potential inhibitor of histamine H1 receptor activity. Osthole also suppresses the secretion of HBV in cells.

Specifications

Chemical Name or Material Osthole
CAS 484-12-8
Color White
Purity Grade Notes Research
Molecular Formula C15H16O3
Quantity 5 g
Synonym Osthol NSC 31868
Solubility Information DMSO : ≥ 100 mg/mL (409.35 mM)
SMILES O=C1C=CC2=CC=C(OC)C(C/C=C(C)\C)=C2O1
Molecular Weight (g/mol) 244.29
Formula Weight 244.29
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.