missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ OSU 6162 hydrochloride
GREENER_CHOICE

Product Code. 18065792
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18065792

Brand: Tocris Bioscience™ 2599/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Dopamine stabilizer

Dopamine stabilizer; lacks high affinity for various neuroreceptors in vitro (Ki values are 447 and 1305 nM for D2 and D3 receptors respectively and > 1 μM for other targets). Exhibits high D2 receptor occupancy in vivo; highly active on dopamine synthesis and turnover. Induces stabilizing effects on psychomotor function in behavioral tests without inducing hypolocomotion or catalepsy.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156907-84-5
Molecular Formula C15H24ClNO2S
Molecular Weight (g/mol) 317.872
InChI Key LEMGVHZVBREXAD-PFEQFJNWSA-N
PubChem CID 9836644
IUPAC Name (3S)-3-(3-methylsulfonylphenyl)-1-propylpiperidine;hydrochloride
SMILES CCCN1CCCC(C1)C2=CC(=CC=C2)S(=O)(=O)C.Cl

Specifications

Quantity 10 mg
Synonym osu 6162 hydrochloride, osu6162 hydrochloride, 3s-3-3-methylsulfonyl phenyl-1-propylpiperidine hydrochloride, pnu-96391; pnu-0096391; osu-6162; s---3-methanesulfonyl-phenyl-1-propyl-piperidine hydrochloride; s,s-3-3-methylsulfonyl phenyl-1-propylpiperidine hydrochloride, 3s-3-3-methanesulfonylphenyl-1-propylpiperidine hydrochloride, unii-v5sbjbujw6, v5sbjbujw6, osu-6162 hydrochloride, osu6162 hydrochloride hplc, 3-3-methanesulfonyl-phenyl-1-propyl-piperidine hydrochloride
Formula Weight 317.87
Percent Purity >98%
Chemical Name or Material OSU 6162 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.