missing translation for 'onlineSavingsMsg'
Learn More

(-)-OSU, TRC

Product Code. 30412530
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412530 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412530 Supplier Toronto Research Chemicals Supplier No. TRCO00616210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (-)-OSU
CAS 146798-66-5
Molecular Formula C15 H23 N O2 S
Quantity 10 mg
Synonym (-)-OSU 6162
InChI Formula InChI=1S/C15H23NO2S/c1-3-9-16-10-5-7-14(12-16)13-6-4-8-15(11-13)19(2,17)18/h4,6,8,11,14H,3,5,7,9-10,12H2,1-2H3/t14-/m1/s1
SMILES CCCN1CCC[C@H](C1)c2cccc(c2)S(=O)(=O)C
IUPAC Name (3S)-3-(3-methylsulfonylphenyl)-1-propylpiperidine
Molecular Weight (g/mol) 281.41
Formula Weight 281.1449
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.