Learn More
uvex OTG Plus Clear SV SAP Overspecs, Black
Product Code. 31751175 Shop All Uvex Products
Change view
Click to view available options
Lens Material:
Polycarbonate
Unit Size:
Each
Product Code. Lens Material Unit Size
31751175 Polycarbonate Each
1 options
Product Code. 31751175 Supplier uvex Supplier No. 9139065
  • ONLINE EXCLUSIVE PRICE
    18.71 €
    17.93 € / Pack of 1
    Save 0.78 €
    4% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed to provide reliable protection and fit comfortably over prescription spectacles.
  • Designed as overspecs that comfortably fit over modern prescription glasses with a large, flat clear SV SAPP polycarbonate lens providing excellent visibility and reliable UV400 eye protection; certified to EN ISO 16321-1 for industrial use and metal-free construction for lightweight comfort.
  • Ergonomic black frame with innovative multi-incline adjustment points and flat, flexible side arms with soft-grip ear-pieces for secure, pressure-free fit on varied facial shapes and head widths.
  • Anatomically shaped nose piece tailored for all-day wear and optimal compatibility with respirators, plus uvex supravision coating for enhanced scratch resistance and durability.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant, Impact-resistant, Anti-fog
Lens Tint None
Product Type Overglasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Adjustable Temples
Autoclavable Not Autoclavable