Learn More
uvex OTG Plus Planet Clear SV EXC Overspecs, Anthracite/Jade
Product Code. 31751169 Shop All Uvex Products
Change view
Click to view available options
Lens Material:
Polycarbonate
Unit Size:
Pack of 8
Product Code. Lens Material Unit Size
31751169 Polycarbonate Pack of 8
1 options
Product Code. 31751169 Supplier uvex Supplier No. 9139295
  • ONLINE EXCLUSIVE PRICE
    177.68 €
    154.27 € / Pack of 8
    Save 23.41 €
    13% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Provide optimum protection and high compatibility with prescription spectacles, featuring a large lens for excellent visibility and comfort.
  • Modern uvex OTG plus overspecs designed for prescription spectacle wearers, featuring an extremely large, flat lens that provides excellent visibility and sufficient space to comfortably wear even large prescription glasses underneath.
  • uvex protecting planet design with a lens made from mass-balanced polycarbonate, a frame partially manufactured from bio-based and recycled materials, plastic-free packaging, 100% recyclable construction, and completely metal-free design.
  • Innovative frame with multi-incline adjustment points allowing a high level of individual customisation to ensure a secure fit for all facial shapes and head widths.
  • Ergonomically shaped, flat flexible side arms with soft-grip ear-pieces deliver a comfortable, secure fit without pressure points, even during extended wear.
  • Anatomically designed nose piece ensures all-day wearing comfort and optimal compatibility with respirators.
  • Certified according to EN ISO 16321-1 for personal eye and face protection in industrial environments, including UV protection filtering.
  • Robust polycarbonate lens with proven uvex supravision coating technology providing permanent anti-fog performance and scratch resistance.
  • UV400 protection offering 100% shielding against hazardous ultraviolet radiation up to 400 nm.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant, Impact-resistant, Anti-fog
Lens Tint None
Product Type Overglasses
Frame Color Green
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Adjustable Temples
Autoclavable Not Autoclavable