missing translation for 'onlineSavingsMsg'
Learn More

Ouabain octahydrate, 95%

Product Code. 15453439
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453439 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15453439 Supplier Thermo Scientific Alfa Aesar Supplier No. J60724.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of sodium-potassium-dependent ATPase | CAS: 11018-89-6 | C29H60O20 | 728.779 g/mol

Ouabain, as well as other endogenous digitalis-like compounds, can down-regulate the expression of 14-3-3 proteins in rat lens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ouabain, as well as other endogenous digitalis-like compounds, can down-regulate the expression of 14-3-3 proteins in rat lens.

Solubility
Soluble in water.

Notes
Protect from light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Specifications

CAS 11018-89-6
Color White
Molecular Formula C29H60O20
MDL Number MFCD00149240
Quantity 1 g
UN Number UN1544
Beilstein 101712
Sensitivity Light sensitive
Merck Index 14,6901
Synonym ouabain octahydrate, strophantine octahydrate, quabain octahydrate, ouabain usp, penta a phenanthren-17-yl-2h-furan-5-one;octahydrate, gamma-strophanthin, 3-1r,3s,5s,8r,9s,10r,11r,13r,14s,17r-1,5,11,14-tetrahydroxy-10-hydroxymethyl-13-methyl-3-2r,3r,4r,5r,6s-3,4,5-trihydroxy-6-methyloxan-2-yl oxy-2,3,4,6,7,8,9,11,12,15,16,17-dodecahydro-1h-cyclo, prestwick_370, acocantherine;g-strophanthin
Solubility Information Soluble in water.
InChI Key TYBARJRCFHUHSN-DMJRSANLSA-N
SMILES CC1C(C(C(C(O1)OC2CC(C3(C4C(CCC3(C2)O)C5(CCC(C5(CC4O)C)C6=CC(=O)OC6)O)CO)O)O)O)O.O.O.O.O.O.O.O.O
IUPAC Name 3-[(1R,3S,5S,8R,9S,10R,11R,13R,14S,17R)-1,5,11,14-tetrahydroxy-10-(hydroxymethyl)-13-methyl-3-[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy-2,3,4,6,7,8,9,11,12,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]-2H-furan-5-one;octahydrate
Molecular Weight (g/mol) 728.779
PubChem CID 6364534
Formula Weight 728.78 (584.66 Anhydrous)
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Ouabain octahydrate
Show More Show Less

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.