missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Ouchterlony Plates

Product Code. 11433966 Shop All MP Biomedicals Products
Change view
Click to view available options
For Use With (Application):
Designed for applications involving antigens present in very low concentrations, prepoured with the wells cut out
Specific and low affinity antisera
Type:
Ouchterlony Plate
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Type Unit Size
11433966 Designed for applications involving antigens present in very low concentrations, prepoured with the wells cut out Ouchterlony Plate Pack of 5
11423966 Specific and low affinity antisera Ouchterlony Plate Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433966 Supplier MP Biomedicals™ Supplier No. 08642781
  • ONLINE EXCLUSIVE PRICE
    699.00 €
    662.55 € / Pack of 5
    Save 36.45 €
    5% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for specific but low affinity antisera that requires properly spaced wells and olyethylene glycol in the agarose for optimal immuno -precipitation.

Specifications

Material Polystyrene
Type Ouchterlony Plate
Well Count 14
For Use With (Application) Designed for applications involving antigens present in very low concentrations, prepoured with the wells cut out
Includes Agarose 1.2% and PEG 5%
Diameter (Metric) 0.3 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.